Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 189864273-189864498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TP63
Entrez Gene ID
8626
Gene Name
tumor protein p63
Gene Aliases
AIS, B(p51A), B(p51B), EEC3, KET, LMS, NBP, OFC8, RHS, SHFM4, TP53CP, TP53L, TP73L, p40, p51, p53CP, p63, p73H, p73L
UniGene
Hs.137569
Gene Chromosome Location
Chr.3: 189597115-189897279 on Build GRCh38
Transcripts
NM_001114978.1 NM_001114979.1 NM_001114980.1 NM_001114981.1 NM_001114982.1 NM_003722.4 XM_005247843.3 XM_005247844.4 XM_011513251.2 XM_011513252.2 XM_017007387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6806659 c.364A>G p.M122V Chr.3: 189864298-189864298 on Build GRCh38
6879548 c.658C>T p.P220S Chr.3: 189864310-189864310 on Build GRCh38
6849518 c.625T>C p.C209R Chr.3: 189864277-189864277 on Build GRCh38
6806690 c.676C>T p.R226C Chr.3: 189864328-189864328 on Build GRCh38
6914257 c.686C>A p.P229H Chr.3: 189864338-189864338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764672477 Chr.3: 189864306-189864306 on Build GRCh38
rs754080925 Chr.3: 189864310-189864310 on Build GRCh38
rs757669482 Chr.3: 189864322-189864322 on Build GRCh38
rs886058221 Chr.3: 189864421-189864421 on Build GRCh38
rs777175930 Chr.3: 189864438-189864438 on Build GRCh38
rs121908836 Chr.3: 189864380-189864380 on Build GRCh38
rs576751351 Chr.3: 189864361-189864361 on Build GRCh38
rs121908835 Chr.3: 189864379-189864379 on Build GRCh38
rs762269643 Chr.3: 189864443-189864443 on Build GRCh38
rs767553568 Chr.3: 189864300-189864300 on Build GRCh38
rs550991181 Chr.3: 189864325-189864325 on Build GRCh38
rs193921145 Chr.3: 189864329-189864329 on Build GRCh38
rs17514215 Chr.3: 189864452-189864452 on Build GRCh38
rs761152678 Chr.3: 189864303-189864303 on Build GRCh38
rs374425727 Chr.3: 189864423-189864423 on Build GRCh38
rs752535926 Chr.3: 189864414-189864414 on Build GRCh38
rs766136969 Chr.3: 189864457-189864457 on Build GRCh38
rs377757904 Chr.3: 189864365-189864365 on Build GRCh38
rs773030906 Chr.3: 189864366-189864366 on Build GRCh38
rs1005174470 Chr.3: 189864381-189864381 on Build GRCh38
rs2276792 Chr.3: 189864460-189864460 on Build GRCh38
rs121908838 Chr.3: 189864349-189864349 on Build GRCh38
rs764465764 Chr.3: 189864429-189864429 on Build GRCh38
rs954483931 Chr.3: 189864455-189864455 on Build GRCh38
rs374833411 Chr.3: 189864326-189864326 on Build GRCh38
rs759801493 Chr.3: 189864298-189864298 on Build GRCh38
rs765633867 Chr.3: 189864444-189864444 on Build GRCh38
rs750875781 Chr.3: 189864451-189864451 on Build GRCh38
rs199686014 Chr.3: 189864363-189864363 on Build GRCh38
rs201617537 Chr.3: 189864377-189864377 on Build GRCh38
rs199727371 Chr.3: 189864351-189864351 on Build GRCh38
rs371524635 Chr.3: 189864402-189864402 on Build GRCh38
rs61732782 Chr.3: 189864330-189864330 on Build GRCh38
rs772334168 Chr.3: 189864405-189864405 on Build GRCh38
rs864621968 Chr.3: 189864392-189864392 on Build GRCh38
rs781222924 Chr.3: 189864345-189864345 on Build GRCh38
rs201466089 Chr.3: 189864340-189864340 on Build GRCh38

View Full Product Details