Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28190982-28191255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLK
Entrez Gene ID
51701
Gene Name
nemo like kinase
Gene Aliases
-
UniGene
Hs.208759
Gene Chromosome Location
Chr.17: 28042647-28205140 on Build GRCh38
Transcripts
NM_016231.4 XM_005257988.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3795364 c.1282G>C p.D428H Chr.17: 28191066-28191066 on Build GRCh38
96071 c.1308A>T p.T436T Chr.17: 28191092-28191092 on Build GRCh38
6726820 c.1298G>A p.R433Q Chr.17: 28191082-28191082 on Build GRCh38
4064878 c.1290G>A p.G430G Chr.17: 28191074-28191074 on Build GRCh38
7081301 c.1240A>G p.K414E Chr.17: 28191024-28191024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764653538 Chr.17: 28191236-28191236 on Build GRCh38
rs55949117 Chr.17: 28191035-28191035 on Build GRCh38
rs755370174 Chr.17: 28191008-28191008 on Build GRCh38
rs774087749 Chr.17: 28191094-28191094 on Build GRCh38
rs759790656 Chr.17: 28191202-28191202 on Build GRCh38
rs137886593 Chr.17: 28191208-28191208 on Build GRCh38
rs763555017 Chr.17: 28191222-28191222 on Build GRCh38
rs753053015 Chr.17: 28191218-28191218 on Build GRCh38
rs760796397 Chr.17: 28191180-28191180 on Build GRCh38
rs753287960 Chr.17: 28191027-28191027 on Build GRCh38
rs35104651 Chr.17: 28191081-28191081 on Build GRCh38
rs755786504 Chr.17: 28191074-28191074 on Build GRCh38
rs769390294 Chr.17: 28191231-28191232 on Build GRCh38
rs34059108 Chr.17: 28191185-28191185 on Build GRCh38
rs780577940 Chr.17: 28191036-28191036 on Build GRCh38
rs548508601 Chr.17: 28191076-28191076 on Build GRCh38
rs760811723 Chr.17: 28191004-28191004 on Build GRCh38
rs748101121 Chr.17: 28191160-28191160 on Build GRCh38
rs146705461 Chr.17: 28191171-28191171 on Build GRCh38
rs140297690 Chr.17: 28191179-28191179 on Build GRCh38
rs776879468 Chr.17: 28191194-28191194 on Build GRCh38
rs768473992 Chr.17: 28191091-28191091 on Build GRCh38
rs562720805 Chr.17: 28191022-28191022 on Build GRCh38
rs754215420 Chr.17: 28191007-28191007 on Build GRCh38
rs959478611 Chr.17: 28191162-28191162 on Build GRCh38
rs34718349 Chr.17: 28191122-28191122 on Build GRCh38
rs765717442 Chr.17: 28191211-28191211 on Build GRCh38
rs745306145 Chr.17: 28191051-28191051 on Build GRCh38
rs766620777 Chr.17: 28191006-28191006 on Build GRCh38

View Full Product Details