Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58422452-58422691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5B2
Entrez Gene ID
390190
Gene Name
olfactory receptor family 5 subfamily B member 2
Gene Aliases
OR11-240, OST073
UniGene
Hs.553753
Gene Chromosome Location
Chr.11: 58422265-58423313 on Build GRCh38
Transcripts
NM_001005566.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
688272 c.729C>A p.F243L Chr.11: 58422533-58422533 on Build GRCh38
3449961 c.606C>T p.S202S Chr.11: 58422656-58422656 on Build GRCh38
688273 c.765C>T p.F255F Chr.11: 58422497-58422497 on Build GRCh38
5897026 c.692G>A p.G231E Chr.11: 58422570-58422570 on Build GRCh38
1218767 c.625C>A p.L209I Chr.11: 58422637-58422637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754958653 Chr.11: 58422657-58422657 on Build GRCh38
rs773690481 Chr.11: 58422647-58422647 on Build GRCh38
rs779236671 Chr.11: 58422502-58422502 on Build GRCh38
rs972126698 Chr.11: 58422538-58422538 on Build GRCh38
rs757719367 Chr.11: 58422495-58422495 on Build GRCh38
rs140988128 Chr.11: 58422480-58422480 on Build GRCh38
rs777537613 Chr.11: 58422600-58422600 on Build GRCh38
rs770681609 Chr.11: 58422525-58422525 on Build GRCh38
rs780927831 Chr.11: 58422583-58422583 on Build GRCh38
rs745329058 Chr.11: 58422620-58422620 on Build GRCh38
rs77603797 Chr.11: 58422631-58422631 on Build GRCh38
rs201076930 Chr.11: 58422559-58422559 on Build GRCh38
rs144933858 Chr.11: 58422506-58422506 on Build GRCh38
rs138755829 Chr.11: 58422513-58422513 on Build GRCh38
rs762200636 Chr.11: 58422549-58422549 on Build GRCh38
rs770464351 Chr.11: 58422611-58422611 on Build GRCh38
rs199619546 Chr.11: 58422615-58422615 on Build GRCh38
rs755222017 Chr.11: 58422540-58422540 on Build GRCh38
rs199846483 Chr.11: 58422532-58422532 on Build GRCh38
rs754228444 Chr.11: 58422494-58422494 on Build GRCh38
rs758492682 Chr.11: 58422510-58422510 on Build GRCh38
rs984934658 Chr.11: 58422596-58422596 on Build GRCh38
rs755117027 Chr.11: 58422576-58422576 on Build GRCh38
rs766777703 Chr.11: 58422662-58422662 on Build GRCh38
rs764699041 Chr.11: 58422478-58422478 on Build GRCh38
rs200368643 Chr.11: 58422567-58422567 on Build GRCh38
rs4298923 Chr.11: 58422663-58422663 on Build GRCh38
rs150284257 Chr.11: 58422521-58422521 on Build GRCh38
rs772207490 Chr.11: 58422603-58422604 on Build GRCh38
rs367805231 Chr.11: 58422652-58422652 on Build GRCh38
rs748100826 Chr.11: 58422585-58422585 on Build GRCh38
rs766471842 Chr.11: 58422656-58422656 on Build GRCh38
rs766673972 Chr.11: 58422569-58422569 on Build GRCh38
rs752843983 Chr.11: 58422660-58422660 on Build GRCh38
rs764455575 Chr.11: 58422536-58422536 on Build GRCh38
rs137992820 Chr.11: 58422556-58422556 on Build GRCh38
rs772360868 Chr.11: 58422540-58422540 on Build GRCh38
rs376113896 Chr.11: 58422530-58422530 on Build GRCh38
rs1035689239 Chr.11: 58422514-58422514 on Build GRCh38
rs773329473 Chr.11: 58422656-58422656 on Build GRCh38
rs202101916 Chr.11: 58422545-58422545 on Build GRCh38
rs140238243 Chr.11: 58422520-58422520 on Build GRCh38
rs751545964 Chr.11: 58422574-58422574 on Build GRCh38
rs777308368 Chr.11: 58422546-58422546 on Build GRCh38
rs926791730 Chr.11: 58422601-58422601 on Build GRCh38
rs549100221 Chr.11: 58422633-58422633 on Build GRCh38
rs371567156 Chr.11: 58422531-58422531 on Build GRCh38
rs750873741 Chr.11: 58422552-58422552 on Build GRCh38
rs749118074 Chr.11: 58422609-58422609 on Build GRCh38
rs1032808163 Chr.11: 58422477-58422477 on Build GRCh38
rs943132397 Chr.11: 58422602-58422602 on Build GRCh38
rs779057590 Chr.11: 58422560-58422560 on Build GRCh38
rs781212701 Chr.11: 58422517-58422517 on Build GRCh38
rs771615510 Chr.11: 58422621-58422621 on Build GRCh38
rs111747013 Chr.11: 58422528-58422528 on Build GRCh38
rs773567025 Chr.11: 58422551-58422551 on Build GRCh38
rs765886541 Chr.11: 58422640-58422640 on Build GRCh38
rs960310914 Chr.11: 58422518-58422518 on Build GRCh38
rs555719372 Chr.11: 58422658-58422658 on Build GRCh38
rs748346476 Chr.11: 58422581-58422582 on Build GRCh38
rs10466659 Chr.11: 58422639-58422639 on Build GRCh38
rs756104711 Chr.11: 58422590-58422590 on Build GRCh38

View Full Product Details