Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121332866-121333103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QRFPR
Entrez Gene ID
84109
Gene Name
pyroglutamylated RFamide peptide receptor
Gene Aliases
AQ27, GPR103, SP9155
UniGene
Hs.368977
Gene Chromosome Location
Chr.4: 121328642-121381029 on Build GRCh38
Transcripts
NM_198179.2 XM_017008693.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
378780 c.582T>A p.Y194* Chr.4: 121333036-121333036 on Build GRCh38
2149989 c.622C>T p.P208S Chr.4: 121332996-121332996 on Build GRCh38
202743 c.718G>T p.E240* Chr.4: 121332900-121332900 on Build GRCh38
3428094 c.562-1G>A p.? Chr.4: 121333057-121333057 on Build GRCh38
4500922 c.576C>A p.F192L Chr.4: 121333042-121333042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759592563 Chr.4: 121333021-121333021 on Build GRCh38
rs139457842 Chr.4: 121332923-121332923 on Build GRCh38
rs746564389 Chr.4: 121332945-121332945 on Build GRCh38
rs762933333 Chr.4: 121332942-121332943 on Build GRCh38
rs756680535 Chr.4: 121333064-121333064 on Build GRCh38
rs1046962572 Chr.4: 121333026-121333026 on Build GRCh38
rs977469806 Chr.4: 121333013-121333013 on Build GRCh38
rs995397062 Chr.4: 121333003-121333003 on Build GRCh38
rs550527843 Chr.4: 121333041-121333041 on Build GRCh38
rs763101064 Chr.4: 121332989-121332989 on Build GRCh38
rs778680995 Chr.4: 121332917-121332917 on Build GRCh38
rs946116238 Chr.4: 121333006-121333006 on Build GRCh38
rs942376930 Chr.4: 121332936-121332936 on Build GRCh38
rs984408619 Chr.4: 121332947-121332947 on Build GRCh38
rs142777322 Chr.4: 121332991-121332991 on Build GRCh38
rs193075383 Chr.4: 121332916-121332916 on Build GRCh38
rs935807134 Chr.4: 121333071-121333071 on Build GRCh38
rs925822475 Chr.4: 121333043-121333043 on Build GRCh38
rs139685422 Chr.4: 121332987-121332987 on Build GRCh38
rs372254772 Chr.4: 121333055-121333055 on Build GRCh38
rs33977434 Chr.4: 121332964-121332964 on Build GRCh38
rs958932028 Chr.4: 121332971-121332971 on Build GRCh38
rs776477870 Chr.4: 121332948-121332948 on Build GRCh38
rs774410992 Chr.4: 121332992-121332992 on Build GRCh38
rs528669363 Chr.4: 121332946-121332946 on Build GRCh38
rs779655405 Chr.4: 121332944-121332944 on Build GRCh38
rs776518040 Chr.4: 121332931-121332931 on Build GRCh38
rs935480016 Chr.4: 121332900-121332900 on Build GRCh38
rs747943209 Chr.4: 121332963-121332963 on Build GRCh38
rs758150585 Chr.4: 121332933-121332933 on Build GRCh38
rs201762984 Chr.4: 121333061-121333061 on Build GRCh38
rs753093066 Chr.4: 121332915-121332915 on Build GRCh38

View Full Product Details