Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160756280-160756521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPM1L
Entrez Gene ID
151742
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1L
Gene Aliases
PP2C-epsilon, PP2CE, PPM1-LIKE
UniGene
Hs.389027
Gene Chromosome Location
Chr.3: 160755532-161078907 on Build GRCh38
Transcripts
NM_139245.3 XM_011512440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6096688 c.86T>C p.L29P Chr.3: 160756394-160756394 on Build GRCh38
3589446 c.108G>A p.W36* Chr.3: 160756416-160756416 on Build GRCh38
384048 c.100G>T p.A34S Chr.3: 160756408-160756408 on Build GRCh38
6753719 c.191C>G p.A64G Chr.3: 160756499-160756499 on Build GRCh38
4634932 c.38G>C p.G13A Chr.3: 160756346-160756346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763587147 Chr.3: 160756437-160756437 on Build GRCh38
rs755160846 Chr.3: 160756366-160756366 on Build GRCh38
rs376368958 Chr.3: 160756449-160756449 on Build GRCh38
rs766592549 Chr.3: 160756358-160756358 on Build GRCh38
rs776870403 Chr.3: 160756322-160756322 on Build GRCh38
rs781420117 Chr.3: 160756368-160756368 on Build GRCh38
rs758793889 Chr.3: 160756372-160756374 on Build GRCh38
rs990492322 Chr.3: 160756429-160756429 on Build GRCh38
rs746715028 Chr.3: 160756419-160756422 on Build GRCh38
rs770336415 Chr.3: 160756434-160756434 on Build GRCh38
rs768283169 Chr.3: 160756321-160756321 on Build GRCh38
rs915122594 Chr.3: 160756472-160756472 on Build GRCh38
rs751619597 Chr.3: 160756365-160756365 on Build GRCh38
rs778656734 Chr.3: 160756390-160756390 on Build GRCh38
rs560428151 Chr.3: 160756493-160756493 on Build GRCh38
rs751009694 Chr.3: 160756347-160756347 on Build GRCh38
rs780350087 Chr.3: 160756375-160756376 on Build GRCh38
rs779539738 Chr.3: 160756409-160756409 on Build GRCh38
rs766926661 Chr.3: 160756441-160756441 on Build GRCh38
rs747726982 Chr.3: 160756433-160756433 on Build GRCh38
rs966514799 Chr.3: 160756395-160756395 on Build GRCh38
rs756280413 Chr.3: 160756474-160756474 on Build GRCh38
rs544863505 Chr.3: 160756393-160756393 on Build GRCh38
rs750147761 Chr.3: 160756489-160756489 on Build GRCh38
rs771953845 Chr.3: 160756405-160756405 on Build GRCh38
rs763536609 Chr.3: 160756350-160756350 on Build GRCh38
rs150937410 Chr.3: 160756384-160756384 on Build GRCh38
rs762293041 Chr.3: 160756332-160756332 on Build GRCh38
rs776415335 Chr.3: 160756416-160756416 on Build GRCh38
rs774931579 Chr.3: 160756299-160756299 on Build GRCh38
rs202235612 Chr.3: 160756479-160756479 on Build GRCh38
rs1032087371 Chr.3: 160756431-160756431 on Build GRCh38
rs139480252 Chr.3: 160756413-160756413 on Build GRCh38
rs752857218 Chr.3: 160756383-160756383 on Build GRCh38
rs746467275 Chr.3: 160756410-160756410 on Build GRCh38
rs369385243 Chr.3: 160756466-160756466 on Build GRCh38
rs765694942 Chr.3: 160756345-160756345 on Build GRCh38
rs760273018 Chr.3: 160756318-160756318 on Build GRCh38
rs150055764 Chr.3: 160756473-160756473 on Build GRCh38

View Full Product Details