Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25983276-25983509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM38
Entrez Gene ID
10475
Gene Name
tripartite motif containing 38
Gene Aliases
RNF15, RORET
UniGene
Hs.584851
Gene Chromosome Location
Chr.6: 25962689-25988867 on Build GRCh38
Transcripts
NM_006355.4 XM_005248799.4 XM_005248800.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
740929 c.1000C>T p.P334S Chr.6: 25983289-25983289 on Build GRCh38
5644582 c.1027A>G p.T343A Chr.6: 25983316-25983316 on Build GRCh38
6808161 c.1162T>C p.C388R Chr.6: 25983451-25983451 on Build GRCh38
6808160 c.1015T>C p.C339R Chr.6: 25983304-25983304 on Build GRCh38
5792675 c.1077G>A p.W359* Chr.6: 25983366-25983366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779359695 Chr.6: 25983351-25983351 on Build GRCh38
rs749276419 Chr.6: 25983413-25983413 on Build GRCh38
rs898995559 Chr.6: 25983343-25983343 on Build GRCh38
rs745440593 Chr.6: 25983375-25983375 on Build GRCh38
rs550157240 Chr.6: 25983467-25983467 on Build GRCh38
rs766598574 Chr.6: 25983485-25983485 on Build GRCh38
rs749278412 Chr.6: 25983396-25983396 on Build GRCh38
rs755260932 Chr.6: 25983322-25983322 on Build GRCh38
rs754224236 Chr.6: 25983313-25983313 on Build GRCh38
rs756936805 Chr.6: 25983360-25983360 on Build GRCh38
rs368108856 Chr.6: 25983361-25983361 on Build GRCh38
rs199895933 Chr.6: 25983420-25983420 on Build GRCh38
rs773408033 Chr.6: 25983478-25983478 on Build GRCh38
rs769693795 Chr.6: 25983376-25983376 on Build GRCh38
rs573211452 Chr.6: 25983464-25983464 on Build GRCh38
rs990034571 Chr.6: 25983447-25983447 on Build GRCh38
rs746583042 Chr.6: 25983352-25983352 on Build GRCh38
rs768283590 Chr.6: 25983418-25983418 on Build GRCh38
rs760879360 Chr.6: 25983484-25983484 on Build GRCh38
rs775442895 Chr.6: 25983395-25983395 on Build GRCh38

View Full Product Details