Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35555134-35555407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RUSC2
Entrez Gene ID
9853
Gene Name
RUN and SH3 domain containing 2
Gene Aliases
Iporin
UniGene
Hs.493796
Gene Chromosome Location
Chr.9: 35487999-35561898 on Build GRCh38
Transcripts
NM_001135999.1 NM_014806.3 NR_052015.2 XM_006716896.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7418401 c.2167T>C p.S723P Chr.9: 35555212-35555212 on Build GRCh38
5942287 c.2236C>T p.P746S Chr.9: 35555281-35555281 on Build GRCh38
6770766 c.2180G>A p.R727H Chr.9: 35555225-35555225 on Build GRCh38
6408320 c.2230C>A p.P744T Chr.9: 35555275-35555275 on Build GRCh38
5977219 c.2354G>A p.S785N Chr.9: 35555399-35555399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018229298 Chr.9: 35555250-35555250 on Build GRCh38
rs776985685 Chr.9: 35555342-35555342 on Build GRCh38
rs746810655 Chr.9: 35555324-35555326 on Build GRCh38
rs777751717 Chr.9: 35555200-35555200 on Build GRCh38
rs141185107 Chr.9: 35555235-35555235 on Build GRCh38
rs143042316 Chr.9: 35555216-35555216 on Build GRCh38
rs182476024 Chr.9: 35555225-35555225 on Build GRCh38
rs766709159 Chr.9: 35555376-35555376 on Build GRCh38
rs148215963 Chr.9: 35555220-35555220 on Build GRCh38
rs555440339 Chr.9: 35555205-35555205 on Build GRCh38
rs780951878 Chr.9: 35555253-35555253 on Build GRCh38
rs201317063 Chr.9: 35555186-35555186 on Build GRCh38
rs1009912648 Chr.9: 35555281-35555281 on Build GRCh38
rs955285995 Chr.9: 35555278-35555278 on Build GRCh38
rs775716958 Chr.9: 35555217-35555217 on Build GRCh38
rs747217604 Chr.9: 35555312-35555312 on Build GRCh38
rs958784489 Chr.9: 35555362-35555362 on Build GRCh38
rs7029864 Chr.9: 35555375-35555375 on Build GRCh38
rs563315271 Chr.9: 35555290-35555290 on Build GRCh38
rs929368608 Chr.9: 35555327-35555327 on Build GRCh38
rs762099808 Chr.9: 35555343-35555343 on Build GRCh38
rs373218246 Chr.9: 35555295-35555295 on Build GRCh38
rs369726144 Chr.9: 35555226-35555226 on Build GRCh38
rs759545337 Chr.9: 35555214-35555214 on Build GRCh38
rs369541476 Chr.9: 35555355-35555355 on Build GRCh38
rs765727835 Chr.9: 35555348-35555348 on Build GRCh38
rs947173608 Chr.9: 35555307-35555307 on Build GRCh38
rs573689022 Chr.9: 35555335-35555335 on Build GRCh38
rs777700666 Chr.9: 35555279-35555279 on Build GRCh38
rs145369360 Chr.9: 35555270-35555270 on Build GRCh38
rs892193726 Chr.9: 35555196-35555196 on Build GRCh38
rs755827653 Chr.9: 35555385-35555385 on Build GRCh38
rs917418097 Chr.9: 35555367-35555367 on Build GRCh38
rs749227407 Chr.9: 35555204-35555204 on Build GRCh38
rs991637509 Chr.9: 35555363-35555363 on Build GRCh38
rs769746982 Chr.9: 35555195-35555195 on Build GRCh38
rs950306578 Chr.9: 35555371-35555371 on Build GRCh38
rs754308469 Chr.9: 35555154-35555154 on Build GRCh38
rs200756109 Chr.9: 35555177-35555177 on Build GRCh38
rs377169613 Chr.9: 35555353-35555353 on Build GRCh38
rs768932260 Chr.9: 35555321-35555321 on Build GRCh38
rs752370555 Chr.9: 35555255-35555255 on Build GRCh38
rs772105000 Chr.9: 35555304-35555304 on Build GRCh38
rs754257661 Chr.9: 35555224-35555224 on Build GRCh38
rs781008748 Chr.9: 35555181-35555181 on Build GRCh38
rs751094554 Chr.9: 35555176-35555176 on Build GRCh38
rs200236775 Chr.9: 35555207-35555207 on Build GRCh38
rs763555777 Chr.9: 35555369-35555369 on Build GRCh38
rs760927545 Chr.9: 35555219-35555219 on Build GRCh38
rs544096152 Chr.9: 35555286-35555286 on Build GRCh38
rs778735033 Chr.9: 35555294-35555294 on Build GRCh38
rs775596143 Chr.9: 35555311-35555311 on Build GRCh38
rs754469316 Chr.9: 35555247-35555247 on Build GRCh38
rs773506860 Chr.9: 35555187-35555187 on Build GRCh38
rs779599994 Chr.9: 35555164-35555164 on Build GRCh38
rs767118616 Chr.9: 35555370-35555370 on Build GRCh38
rs532275583 Chr.9: 35555326-35555326 on Build GRCh38

View Full Product Details