Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 173604725-173604998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLGN1
Entrez Gene ID
22871
Gene Name
neuroligin 1
Gene Aliases
NL1
UniGene
Hs.478289
Gene Chromosome Location
Chr.3: 173395899-174291287 on Build GRCh38
Transcripts
NM_014932.3 XM_005247234.2 XM_005247235.3 XM_005247236.2 XM_006713540.3 XM_011512551.2 XM_011512552.2 XM_017005888.1 XM_017005889.1 XM_017005890.1 XM_017005891.1 XM_017005892.1 XM_017005893.1 XM_017005894.1 XM_017005895.1 XM_017005896.1 XM_017005897.1 XM_017005898.1 XM_017005899.1 XM_017005900.1 XM_017005901.1 XM_017005902.1 XM_017005903.1 XM_017005904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4531928 c.183G>A p.K61K Chr.3: 173604781-173604781 on Build GRCh38
5994853 c.327T>C p.N109N Chr.3: 173604925-173604925 on Build GRCh38
5761433 c.332C>T p.T111I Chr.3: 173604930-173604930 on Build GRCh38
4115341 c.371G>A p.G124D Chr.3: 173604969-173604969 on Build GRCh38
5886290 c.271G>A p.G91R Chr.3: 173604869-173604869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760812996 Chr.3: 173604897-173604897 on Build GRCh38
rs749776633 Chr.3: 173604876-173604876 on Build GRCh38
rs1052981284 Chr.3: 173604872-173604872 on Build GRCh38
rs747696776 Chr.3: 173604863-173604863 on Build GRCh38
rs201769872 Chr.3: 173604933-173604933 on Build GRCh38
rs778406192 Chr.3: 173604871-173604871 on Build GRCh38
rs759229363 Chr.3: 173604943-173604943 on Build GRCh38
rs772330006 Chr.3: 173604895-173604895 on Build GRCh38
rs768630828 Chr.3: 173604902-173604902 on Build GRCh38
rs759760720 Chr.3: 173604802-173604802 on Build GRCh38
rs1021469652 Chr.3: 173604823-173604823 on Build GRCh38
rs773132929 Chr.3: 173604905-173604905 on Build GRCh38
rs774580240 Chr.3: 173604800-173604800 on Build GRCh38
rs554696421 Chr.3: 173604775-173604775 on Build GRCh38
rs552792410 Chr.3: 173604888-173604888 on Build GRCh38
rs966805126 Chr.3: 173604861-173604861 on Build GRCh38
rs758153927 Chr.3: 173604835-173604835 on Build GRCh38
rs746042193 Chr.3: 173604893-173604893 on Build GRCh38
rs771049032 Chr.3: 173604794-173604794 on Build GRCh38
rs766147986 Chr.3: 173604938-173604938 on Build GRCh38
rs997502640 Chr.3: 173604817-173604817 on Build GRCh38
rs754696590 Chr.3: 173604850-173604850 on Build GRCh38
rs751215436 Chr.3: 173604845-173604845 on Build GRCh38
rs996071901 Chr.3: 173604918-173604918 on Build GRCh38
rs761718633 Chr.3: 173604818-173604818 on Build GRCh38
rs770469708 Chr.3: 173604765-173604765 on Build GRCh38
rs748614817 Chr.3: 173604752-173604752 on Build GRCh38
rs1007429131 Chr.3: 173604757-173604757 on Build GRCh38
rs1030178857 Chr.3: 173604935-173604935 on Build GRCh38
rs141555720 Chr.3: 173604853-173604853 on Build GRCh38
rs780824820 Chr.3: 173604860-173604860 on Build GRCh38
rs374685079 Chr.3: 173604878-173604878 on Build GRCh38
rs765261134 Chr.3: 173604821-173604821 on Build GRCh38
rs371254567 Chr.3: 173604825-173604825 on Build GRCh38
rs145279213 Chr.3: 173604867-173604867 on Build GRCh38
rs767547807 Chr.3: 173604804-173604804 on Build GRCh38
rs200519167 Chr.3: 173604963-173604963 on Build GRCh38
rs935435410 Chr.3: 173604870-173604870 on Build GRCh38
rs752239356 Chr.3: 173604959-173604959 on Build GRCh38
rs775571610 Chr.3: 173604807-173604807 on Build GRCh38
rs773413102 Chr.3: 173604774-173604774 on Build GRCh38
rs140900040 Chr.3: 173604896-173604896 on Build GRCh38
rs750181718 Chr.3: 173604830-173604830 on Build GRCh38
rs777154584 Chr.3: 173604966-173604966 on Build GRCh38
rs150137049 Chr.3: 173604946-173604946 on Build GRCh38
rs751446277 Chr.3: 173604941-173604941 on Build GRCh38
rs766090131 Chr.3: 173604837-173604837 on Build GRCh38
rs1046569031 Chr.3: 173604859-173604859 on Build GRCh38

View Full Product Details