Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100024067-100024340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZKSCAN1
Entrez Gene ID
7586
Gene Name
zinc finger with KRAB and SCAN domains 1
Gene Aliases
9130423L19Rik, KOX18, PHZ-37, ZNF139, ZNF36, ZSCAN33
UniGene
Hs.615360
Gene Chromosome Location
Chr.7: 100015572-100045523 on Build GRCh38
Transcripts
NM_001287054.1 NM_001287055.1 NM_003439.2 XM_005250565.4 XM_011516559.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3883905 c.554G>A p.R185Q Chr.7: 100024281-100024281 on Build GRCh38
3412577 c.466G>A p.G156R Chr.7: 100024193-100024193 on Build GRCh38
1699013 c.551C>T p.S184F Chr.7: 100024278-100024278 on Build GRCh38
5710772 c.580G>A p.A194T Chr.7: 100024307-100024307 on Build GRCh38
5627266 c.493G>A p.E165K Chr.7: 100024220-100024220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761448127 Chr.7: 100024107-100024107 on Build GRCh38
rs868197329 Chr.7: 100024178-100024178 on Build GRCh38
rs1046907956 Chr.7: 100024237-100024237 on Build GRCh38
rs781502503 Chr.7: 100024227-100024227 on Build GRCh38
rs770288802 Chr.7: 100024251-100024251 on Build GRCh38
rs752517745 Chr.7: 100024281-100024281 on Build GRCh38
rs201057645 Chr.7: 100024289-100024289 on Build GRCh38
rs762316729 Chr.7: 100024132-100024132 on Build GRCh38
rs915915274 Chr.7: 100024150-100024150 on Build GRCh38
rs1042036628 Chr.7: 100024095-100024095 on Build GRCh38
rs765944933 Chr.7: 100024157-100024157 on Build GRCh38
rs142959093 Chr.7: 100024214-100024214 on Build GRCh38
rs867920124 Chr.7: 100024198-100024198 on Build GRCh38
rs923732517 Chr.7: 100024135-100024135 on Build GRCh38
rs933703997 Chr.7: 100024138-100024138 on Build GRCh38
rs878961968 Chr.7: 100024113-100024113 on Build GRCh38
rs748558829 Chr.7: 100024228-100024228 on Build GRCh38
rs773967457 Chr.7: 100024264-100024264 on Build GRCh38
rs1007180800 Chr.7: 100024196-100024196 on Build GRCh38
rs759540374 Chr.7: 100024275-100024275 on Build GRCh38
rs762671026 Chr.7: 100024247-100024247 on Build GRCh38
rs145281775 Chr.7: 100024171-100024171 on Build GRCh38
rs751386566 Chr.7: 100024159-100024159 on Build GRCh38
rs376585197 Chr.7: 100024108-100024108 on Build GRCh38
rs780192057 Chr.7: 100024199-100024199 on Build GRCh38
rs750680760 Chr.7: 100024195-100024195 on Build GRCh38
rs758664117 Chr.7: 100024291-100024291 on Build GRCh38
rs980715811 Chr.7: 100024181-100024181 on Build GRCh38
rs202154703 Chr.7: 100024290-100024290 on Build GRCh38
rs148037051 Chr.7: 100024305-100024305 on Build GRCh38
rs559885441 Chr.7: 100024141-100024141 on Build GRCh38
rs149163835 Chr.7: 100024187-100024187 on Build GRCh38
rs762080657 Chr.7: 100024276-100024276 on Build GRCh38
rs767405218 Chr.7: 100024186-100024186 on Build GRCh38
rs148297517 Chr.7: 100024090-100024090 on Build GRCh38
rs374057748 Chr.7: 100024129-100024129 on Build GRCh38
rs760499332 Chr.7: 100024285-100024285 on Build GRCh38
rs774416307 Chr.7: 100024246-100024246 on Build GRCh38
rs770161589 Chr.7: 100024238-100024238 on Build GRCh38
rs769212492 Chr.7: 100024222-100024222 on Build GRCh38
rs143265277 Chr.7: 100024191-100024191 on Build GRCh38

View Full Product Details