Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 10044195-10044468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKEF1
Entrez Gene ID
63926
Gene Name
ankyrin repeat and EF-hand domain containing 1
Gene Aliases
ANKRD5
UniGene
Hs.70903
Gene Chromosome Location
Chr.20: 10034999-10056764 on Build GRCh38
Transcripts
NM_001303472.1 NM_022096.5 NM_198798.2 XM_005260792.2 NR_040710.1

Gene Symbol
SNAP25-AS1
Entrez Gene ID
100131208
Gene Name
SNAP25 antisense RNA 1
Gene Aliases
-
UniGene
Hs.658178
Gene Chromosome Location
Chr.20: 10023812-10219506 on Build GRCh38
Transcripts
NM_001303472.1 NM_022096.5 NM_198798.2 XM_005260792.2 NR_040710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661928 c.575C>T p.A192V Chr.20: 10044422-10044422 on Build GRCh38
172021 c.604C>T p.R202* Chr.20: 10044451-10044451 on Build GRCh38
6599990 c.579A>G p.S193S Chr.20: 10044426-10044426 on Build GRCh38
4661929 c.576G>A p.A192A Chr.20: 10044423-10044423 on Build GRCh38
6092671 c.588G>T p.G196G Chr.20: 10044435-10044435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745529728 Chr.20: 10044415-10044415 on Build GRCh38
rs1032095072 Chr.20: 10044273-10044273 on Build GRCh38
rs139757926 Chr.20: 10044403-10044403 on Build GRCh38
rs1000380410 Chr.20: 10044277-10044277 on Build GRCh38
rs774166530 Chr.20: 10044387-10044387 on Build GRCh38
rs142822021 Chr.20: 10044423-10044423 on Build GRCh38
rs754980557 Chr.20: 10044394-10044394 on Build GRCh38
rs1007800705 Chr.20: 10044339-10044339 on Build GRCh38
rs780834065 Chr.20: 10044413-10044413 on Build GRCh38
rs386812407 Chr.20: 10044302-10044303 on Build GRCh38
rs6039662 Chr.20: 10044302-10044302 on Build GRCh38
rs768614213 Chr.20: 10044436-10044436 on Build GRCh38
rs766610346 Chr.20: 10044388-10044388 on Build GRCh38
rs931320247 Chr.20: 10044439-10044439 on Build GRCh38
rs138914733 Chr.20: 10044258-10044258 on Build GRCh38
rs117704926 Chr.20: 10044355-10044355 on Build GRCh38
rs529316906 Chr.20: 10044231-10044231 on Build GRCh38
rs765309622 Chr.20: 10044396-10044396 on Build GRCh38
rs201790922 Chr.20: 10044421-10044421 on Build GRCh38
rs6039663 Chr.20: 10044303-10044303 on Build GRCh38
rs754074424 Chr.20: 10044389-10044389 on Build GRCh38
rs1025604732 Chr.20: 10044246-10044246 on Build GRCh38
rs555309678 Chr.20: 10044426-10044426 on Build GRCh38
rs748848277 Chr.20: 10044433-10044433 on Build GRCh38
rs1055561443 Chr.20: 10044441-10044441 on Build GRCh38
rs867301083 Chr.20: 10044392-10044392 on Build GRCh38
rs763943502 Chr.20: 10044367-10044367 on Build GRCh38
rs753150832 Chr.20: 10044398-10044398 on Build GRCh38
rs966440536 Chr.20: 10044251-10044251 on Build GRCh38
rs957665836 Chr.20: 10044278-10044278 on Build GRCh38
rs796354028 Chr.20: 10044307-10044307 on Build GRCh38
rs570339688 Chr.20: 10044344-10044344 on Build GRCh38
rs569271468 Chr.20: 10044283-10044283 on Build GRCh38
rs183548235 Chr.20: 10044224-10044224 on Build GRCh38
rs367573452 Chr.20: 10044418-10044418 on Build GRCh38
rs757633997 Chr.20: 10044410-10044410 on Build GRCh38
rs145076695 Chr.20: 10044404-10044404 on Build GRCh38

View Full Product Details