Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6409968-6410233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAGLB
Entrez Gene ID
221955
Gene Name
diacylglycerol lipase beta
Gene Aliases
DAGLBETA, KCCR13L
UniGene
Hs.487498
Gene Chromosome Location
Chr.7: 6409116-6448012 on Build GRCh38
Transcripts
NM_001142936.1 NM_139179.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
118994 c.1820+2T>C p.? Chr.7: 6410128-6410128 on Build GRCh38
3412231 c.1888C>T p.L630F Chr.7: 6409968-6409968 on Build GRCh38
7293554 c.1841C>T p.A614V Chr.7: 6410015-6410015 on Build GRCh38
237271 c.1820+9G>A p.? Chr.7: 6410121-6410121 on Build GRCh38
7363678 c.1787T>C p.I596T Chr.7: 6410163-6410163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753682676 Chr.7: 6410081-6410082 on Build GRCh38
rs757677803 Chr.7: 6410003-6410003 on Build GRCh38
rs748783259 Chr.7: 6410028-6410028 on Build GRCh38
rs368005165 Chr.7: 6410199-6410199 on Build GRCh38
rs1054966238 Chr.7: 6410141-6410141 on Build GRCh38
rs746719305 Chr.7: 6410186-6410186 on Build GRCh38
rs368426784 Chr.7: 6410122-6410122 on Build GRCh38
rs746671332 Chr.7: 6410183-6410184 on Build GRCh38
rs547236199 Chr.7: 6410045-6410045 on Build GRCh38
rs117763067 Chr.7: 6410040-6410040 on Build GRCh38
rs538724319 Chr.7: 6410092-6410092 on Build GRCh38
rs372922766 Chr.7: 6410066-6410066 on Build GRCh38
rs868281554 Chr.7: 6410212-6410212 on Build GRCh38
rs371193609 Chr.7: 6410118-6410118 on Build GRCh38
rs767303133 Chr.7: 6410121-6410121 on Build GRCh38
rs777360450 Chr.7: 6410125-6410125 on Build GRCh38
rs747702778 Chr.7: 6410111-6410119 on Build GRCh38
rs755849106 Chr.7: 6410013-6410020 on Build GRCh38
rs760967486 Chr.7: 6410167-6410167 on Build GRCh38
rs751896933 Chr.7: 6409994-6409994 on Build GRCh38
rs1034871158 Chr.7: 6410187-6410187 on Build GRCh38
rs774502686 Chr.7: 6410147-6410147 on Build GRCh38
rs746629502 Chr.7: 6410126-6410126 on Build GRCh38
rs372289619 Chr.7: 6410124-6410124 on Build GRCh38
rs766835420 Chr.7: 6410170-6410170 on Build GRCh38
rs149455276 Chr.7: 6410185-6410185 on Build GRCh38
rs764046268 Chr.7: 6410057-6410057 on Build GRCh38
rs1028703911 Chr.7: 6410134-6410134 on Build GRCh38
rs755525042 Chr.7: 6410183-6410183 on Build GRCh38
rs760614797 Chr.7: 6410050-6410050 on Build GRCh38
rs188076040 Chr.7: 6410065-6410065 on Build GRCh38
rs766251087 Chr.7: 6410052-6410052 on Build GRCh38
rs767674693 Chr.7: 6410155-6410155 on Build GRCh38
rs751854647 Chr.7: 6410058-6410058 on Build GRCh38
rs765616195 Chr.7: 6410184-6410184 on Build GRCh38
rs749727716 Chr.7: 6410068-6410068 on Build GRCh38
rs770882845 Chr.7: 6410087-6410087 on Build GRCh38
rs758121991 Chr.7: 6410118-6410122 on Build GRCh38
rs762129170 Chr.7: 6410113-6410113 on Build GRCh38
rs866999970 Chr.7: 6410146-6410146 on Build GRCh38
rs146258893 Chr.7: 6410085-6410085 on Build GRCh38
rs138947170 Chr.7: 6410009-6410009 on Build GRCh38
rs904683502 Chr.7: 6410110-6410110 on Build GRCh38
rs768224382 Chr.7: 6410029-6410029 on Build GRCh38
rs543787333 Chr.7: 6410131-6410131 on Build GRCh38
rs756853830 Chr.7: 6410192-6410192 on Build GRCh38
rs139753251 Chr.7: 6410163-6410163 on Build GRCh38
rs750208748 Chr.7: 6410012-6410019 on Build GRCh38
rs761351765 Chr.7: 6410036-6410036 on Build GRCh38
rs767789487 Chr.7: 6410116-6410116 on Build GRCh38
rs781044427 Chr.7: 6410195-6410195 on Build GRCh38
rs769550995 Chr.7: 6410136-6410136 on Build GRCh38
rs754965882 Chr.7: 6410102-6410110 on Build GRCh38
rs920207493 Chr.7: 6410076-6410076 on Build GRCh38
rs974278622 Chr.7: 6410069-6410069 on Build GRCh38
rs549260966 Chr.7: 6410056-6410056 on Build GRCh38
rs747602404 Chr.7: 6410123-6410123 on Build GRCh38
rs778264977 Chr.7: 6410101-6410102 on Build GRCh38
rs778275119 Chr.7: 6410081-6410081 on Build GRCh38
rs375425034 Chr.7: 6410119-6410119 on Build GRCh38
rs777624890 Chr.7: 6410179-6410180 on Build GRCh38
rs867702263 Chr.7: 6410140-6410140 on Build GRCh38
rs754086904 Chr.7: 6410171-6410171 on Build GRCh38
rs141322626 Chr.7: 6410021-6410021 on Build GRCh38
rs557993902 Chr.7: 6410101-6410101 on Build GRCh38
rs771915125 Chr.7: 6410038-6410038 on Build GRCh38
rs960883296 Chr.7: 6410189-6410189 on Build GRCh38
rs201911372 Chr.7: 6410130-6410130 on Build GRCh38
rs368654531 Chr.7: 6410088-6410088 on Build GRCh38
rs370327730 Chr.7: 6410077-6410077 on Build GRCh38
rs954203348 Chr.7: 6410084-6410084 on Build GRCh38
rs774094907 Chr.7: 6410030-6410030 on Build GRCh38
rs377478824 Chr.7: 6410067-6410067 on Build GRCh38
rs371012241 Chr.7: 6410004-6410004 on Build GRCh38
rs148484155 Chr.7: 6410135-6410135 on Build GRCh38
rs200924183 Chr.7: 6410086-6410086 on Build GRCh38
rs896201413 Chr.7: 6410159-6410159 on Build GRCh38
rs746434139 Chr.7: 6410005-6410005 on Build GRCh38
rs774289572 Chr.7: 6410111-6410111 on Build GRCh38
rs747903762 Chr.7: 6410082-6410082 on Build GRCh38
rs145984096 Chr.7: 6410202-6410202 on Build GRCh38

View Full Product Details