Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 15074666-15074913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCB
Entrez Gene ID
797
Gene Name
calcitonin related polypeptide beta
Gene Aliases
CALC2, CGRP-II, CGRP2
UniGene
Hs.534305
Gene Chromosome Location
Chr.11: 15073600-15078631 on Build GRCh38
Transcripts
NM_000728.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1352848 c.54C>T p.Y18Y Chr.11: 15074772-15074772 on Build GRCh38
3445664 c.11G>A p.R4Q Chr.11: 15074729-15074729 on Build GRCh38
5739182 c.74C>A p.A25E Chr.11: 15074792-15074792 on Build GRCh38
1746151 c.36C>T p.L12L Chr.11: 15074754-15074754 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767618344 Chr.11: 15074852-15074852 on Build GRCh38
rs761441692 Chr.11: 15074726-15074726 on Build GRCh38
rs200217522 Chr.11: 15074828-15074828 on Build GRCh38
rs766953127 Chr.11: 15074739-15074739 on Build GRCh38
rs376322094 Chr.11: 15074787-15074787 on Build GRCh38
rs776916764 Chr.11: 15074853-15074857 on Build GRCh38
rs780002303 Chr.11: 15074694-15074694 on Build GRCh38
rs369719079 Chr.11: 15074789-15074789 on Build GRCh38
rs1540139 Chr.11: 15074873-15074873 on Build GRCh38
rs1001276587 Chr.11: 15074692-15074692 on Build GRCh38
rs771935482 Chr.11: 15074693-15074693 on Build GRCh38
rs761125737 Chr.11: 15074737-15074738 on Build GRCh38
rs773557900 Chr.11: 15074728-15074729 on Build GRCh38
rs980235610 Chr.11: 15074776-15074776 on Build GRCh38
rs768209569 Chr.11: 15074812-15074812 on Build GRCh38
rs770782015 Chr.11: 15074719-15074719 on Build GRCh38
rs139794988 Chr.11: 15074778-15074778 on Build GRCh38
rs1055368526 Chr.11: 15074727-15074727 on Build GRCh38
rs746477876 Chr.11: 15074703-15074703 on Build GRCh38
rs746992399 Chr.11: 15074803-15074803 on Build GRCh38
rs780627046 Chr.11: 15074817-15074817 on Build GRCh38
rs763415957 Chr.11: 15074738-15074738 on Build GRCh38
rs774965950 Chr.11: 15074848-15074848 on Build GRCh38
rs776318498 Chr.11: 15074723-15074723 on Build GRCh38
rs771470992 Chr.11: 15074847-15074847 on Build GRCh38
rs533078689 Chr.11: 15074882-15074882 on Build GRCh38
rs768103547 Chr.11: 15074710-15074710 on Build GRCh38
rs773655356 Chr.11: 15074736-15074736 on Build GRCh38
rs760025848 Chr.11: 15074851-15074851 on Build GRCh38
rs369534959 Chr.11: 15074829-15074829 on Build GRCh38
rs763360487 Chr.11: 15074831-15074831 on Build GRCh38
rs75461927 Chr.11: 15074890-15074890 on Build GRCh38
rs1051124289 Chr.11: 15074894-15074894 on Build GRCh38
rs752957836 Chr.11: 15074768-15074768 on Build GRCh38
rs766907604 Chr.11: 15074766-15074766 on Build GRCh38
rs767532661 Chr.11: 15074760-15074760 on Build GRCh38
rs529327624 Chr.11: 15074741-15074741 on Build GRCh38
rs181580047 Chr.11: 15074707-15074707 on Build GRCh38
rs756345561 Chr.11: 15074771-15074771 on Build GRCh38
rs770182897 Chr.11: 15074729-15074729 on Build GRCh38
rs779948872 Chr.11: 15074795-15074795 on Build GRCh38
rs759637036 Chr.11: 15074754-15074754 on Build GRCh38

View Full Product Details