Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10801596-10801855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4G2
Entrez Gene ID
1982
Gene Name
eukaryotic translation initiation factor 4 gamma 2
Gene Aliases
AAG1, DAP5, NAT1, P97
UniGene
Hs.183684
Gene Chromosome Location
Chr.11: 10797046-10809035 on Build GRCh38
Transcripts
NM_001042559.2 NM_001172705.1 NM_001418.3 NR_004403.1

Gene Symbol
SNORD97
Entrez Gene ID
692223
Gene Name
small nucleolar RNA, C/D box 97
Gene Aliases
U97
UniGene
Hs.723086
Gene Chromosome Location
Chr.11: 10801467-10801608 on Build GRCh38
Transcripts
NM_001042559.2 NM_001172705.1 NM_001418.3 NR_004403.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
922758 c.1326G>C p.Q442H Chr.11: 10801748-10801748 on Build GRCh38
922757 c.1361C>T p.S454L Chr.11: 10801713-10801713 on Build GRCh38
7125236 c.1378C>T p.R460W Chr.11: 10801696-10801696 on Build GRCh38
1251095 c.1378C>A p.R460R Chr.11: 10801696-10801696 on Build GRCh38
4780378 c.1339T>C p.L447L Chr.11: 10801735-10801735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763580658 Chr.11: 10801788-10801788 on Build GRCh38
rs972069289 Chr.11: 10801707-10801707 on Build GRCh38
rs761551132 Chr.11: 10801624-10801624 on Build GRCh38
rs930741813 Chr.11: 10801720-10801720 on Build GRCh38
rs944691822 Chr.11: 10801656-10801656 on Build GRCh38
rs768504147 Chr.11: 10801702-10801702 on Build GRCh38
rs763083606 Chr.11: 10801730-10801730 on Build GRCh38
rs751605803 Chr.11: 10801735-10801735 on Build GRCh38
rs745490693 Chr.11: 10801654-10801654 on Build GRCh38
rs375199297 Chr.11: 10801796-10801796 on Build GRCh38
rs769499738 Chr.11: 10801640-10801646 on Build GRCh38
rs374179465 Chr.11: 10801629-10801629 on Build GRCh38
rs753222754 Chr.11: 10801739-10801739 on Build GRCh38
rs764998559 Chr.11: 10801712-10801712 on Build GRCh38
rs948904058 Chr.11: 10801773-10801773 on Build GRCh38
rs750615306 Chr.11: 10801632-10801632 on Build GRCh38
rs745580672 Chr.11: 10801783-10801783 on Build GRCh38
rs551579662 Chr.11: 10801827-10801827 on Build GRCh38
rs199903099 Chr.11: 10801816-10801816 on Build GRCh38
rs200053038 Chr.11: 10801637-10801637 on Build GRCh38
rs766532000 Chr.11: 10801821-10801821 on Build GRCh38
rs370394820 Chr.11: 10801648-10801648 on Build GRCh38
rs878881033 Chr.11: 10801777-10801777 on Build GRCh38
rs762579598 Chr.11: 10801776-10801777 on Build GRCh38
rs1031772072 Chr.11: 10801817-10801817 on Build GRCh38
rs779696469 Chr.11: 10801791-10801791 on Build GRCh38
rs775150822 Chr.11: 10801775-10801776 on Build GRCh38
rs758523077 Chr.11: 10801638-10801638 on Build GRCh38
rs770242475 Chr.11: 10801649-10801649 on Build GRCh38
rs772996357 Chr.11: 10801815-10801815 on Build GRCh38
rs199909225 Chr.11: 10801631-10801631 on Build GRCh38
rs773898466 Chr.11: 10801790-10801793 on Build GRCh38
rs1045994345 Chr.11: 10801776-10801776 on Build GRCh38
rs753651240 Chr.11: 10801623-10801623 on Build GRCh38
rs75174859 Chr.11: 10801717-10801717 on Build GRCh38
rs768169919 Chr.11: 10801736-10801736 on Build GRCh38
rs771668658 Chr.11: 10801675-10801675 on Build GRCh38
rs774854761 Chr.11: 10801689-10801689 on Build GRCh38
rs766557150 Chr.11: 10801732-10801732 on Build GRCh38
rs778266008 Chr.11: 10801781-10801781 on Build GRCh38
rs202215006 Chr.11: 10801704-10801704 on Build GRCh38
rs185328005 Chr.11: 10801709-10801709 on Build GRCh38
rs367756091 Chr.11: 10801646-10801646 on Build GRCh38
rs778746414 Chr.11: 10801652-10801652 on Build GRCh38
rs768184639 Chr.11: 10801795-10801795 on Build GRCh38
rs148204209 Chr.11: 10801772-10801772 on Build GRCh38
rs746458672 Chr.11: 10801696-10801696 on Build GRCh38
rs769644990 Chr.11: 10801812-10801812 on Build GRCh38
rs767221726 Chr.11: 10801789-10801789 on Build GRCh38
rs763200907 Chr.11: 10801636-10801636 on Build GRCh38
rs779987229 Chr.11: 10801642-10801642 on Build GRCh38
rs879242348 Chr.11: 10801693-10801693 on Build GRCh38
rs776514344 Chr.11: 10801705-10801705 on Build GRCh38
rs773483249 Chr.11: 10801713-10801713 on Build GRCh38
rs756593971 Chr.11: 10801755-10801755 on Build GRCh38

View Full Product Details