Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7218054-7218311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADVL
Entrez Gene ID
37
Gene Name
acyl-CoA dehydrogenase, very long chain
Gene Aliases
ACAD6, LCACD, VLCAD
UniGene
Hs.437178
Gene Chromosome Location
Chr.17: 7217125-7225267 on Build GRCh38
Transcripts
NM_001270447.1 NM_001321074.1 NM_001365.4 NR_135527.1 XM_011523698.1 XM_011523699.2

Gene Symbol
DLG4
Entrez Gene ID
1742
Gene Name
discs large MAGUK scaffold protein 4
Gene Aliases
PSD95, SAP-90, SAP90
UniGene
Hs.463928
Gene Chromosome Location
Chr.17: 7189890-7220050 on Build GRCh38
Transcripts
NM_001270447.1 NM_001321074.1 NM_001365.4 NR_135527.1 XM_011523698.1 XM_011523699.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7148363 c.127G>T p.D43Y Chr.17: 7218273-7218273 on Build GRCh38
5170321 c.129C>A p.D43E Chr.17: 7218271-7218271 on Build GRCh38
6556081 c.156T>A p.S52R Chr.17: 7218244-7218244 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773831815 Chr.17: 7218122-7218123 on Build GRCh38
rs759500299 Chr.17: 7218224-7218224 on Build GRCh38
rs374655022 Chr.17: 7218187-7218187 on Build GRCh38
rs776540362 Chr.17: 7218226-7218226 on Build GRCh38
rs372731000 Chr.17: 7218279-7218279 on Build GRCh38
rs577474917 Chr.17: 7218221-7218221 on Build GRCh38
rs969652365 Chr.17: 7218085-7218086 on Build GRCh38
rs780212257 Chr.17: 7218245-7218246 on Build GRCh38
rs764063261 Chr.17: 7218225-7218225 on Build GRCh38
rs951378823 Chr.17: 7218273-7218273 on Build GRCh38
rs758092695 Chr.17: 7218256-7218256 on Build GRCh38
rs41283399 Chr.17: 7218220-7218220 on Build GRCh38
rs761667426 Chr.17: 7218235-7218235 on Build GRCh38
rs1035239537 Chr.17: 7218110-7218110 on Build GRCh38
rs754522496 Chr.17: 7218275-7218275 on Build GRCh38
rs750130441 Chr.17: 7218245-7218245 on Build GRCh38
rs960172345 Chr.17: 7218211-7218211 on Build GRCh38
rs748668101 Chr.17: 7218283-7218283 on Build GRCh38
rs768526024 Chr.17: 7218277-7218277 on Build GRCh38
rs2736880 Chr.17: 7218263-7218263 on Build GRCh38
rs765908419 Chr.17: 7218257-7218257 on Build GRCh38
rs771980584 Chr.17: 7218099-7218099 on Build GRCh38
rs765023240 Chr.17: 7218240-7218240 on Build GRCh38
rs375520253 Chr.17: 7218208-7218208 on Build GRCh38
rs986452193 Chr.17: 7218166-7218166 on Build GRCh38

View Full Product Details