Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116635787-116636030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZUFSP
Entrez Gene ID
221302
Gene Name
zinc finger with UFM1 specific peptidase domain
Gene Aliases
C6orf113, dJ412I7.3
UniGene
Hs.29857
Gene Chromosome Location
Chr.6: 116635618-116668810 on Build GRCh38
Transcripts
NM_145062.2 XM_011535565.2 XM_011535567.1 XM_017010394.1 XM_017010395.1 XM_017010396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6172075 c.1736G>A p.*579* Chr.6: 116635833-116635833 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763524620 Chr.6: 116635879-116635879 on Build GRCh38
rs767933348 Chr.6: 116635909-116635909 on Build GRCh38
rs1005640399 Chr.6: 116635988-116635988 on Build GRCh38
rs765360043 Chr.6: 116635871-116635871 on Build GRCh38
rs78669060 Chr.6: 116635852-116635852 on Build GRCh38
rs1044135630 Chr.6: 116635965-116635965 on Build GRCh38
rs911399543 Chr.6: 116635826-116635826 on Build GRCh38
rs942917849 Chr.6: 116635831-116635831 on Build GRCh38
rs200868601 Chr.6: 116635847-116635847 on Build GRCh38
rs2295693 Chr.6: 116635981-116635981 on Build GRCh38
rs200837760 Chr.6: 116635922-116635922 on Build GRCh38
rs150822257 Chr.6: 116635837-116635837 on Build GRCh38
rs371644401 Chr.6: 116635989-116635989 on Build GRCh38
rs773475871 Chr.6: 116635876-116635876 on Build GRCh38
rs946681526 Chr.6: 116635935-116635935 on Build GRCh38
rs751627186 Chr.6: 116635846-116635846 on Build GRCh38
rs754063348 Chr.6: 116635883-116635887 on Build GRCh38
rs755244530 Chr.6: 116635902-116635902 on Build GRCh38
rs994562104 Chr.6: 116635918-116635918 on Build GRCh38
rs1039255794 Chr.6: 116635866-116635866 on Build GRCh38
rs1047107033 Chr.6: 116635980-116635980 on Build GRCh38
rs751942748 Chr.6: 116635895-116635895 on Build GRCh38
rs755134093 Chr.6: 116635895-116635896 on Build GRCh38
rs766864615 Chr.6: 116635888-116635888 on Build GRCh38
rs779018676 Chr.6: 116635903-116635908 on Build GRCh38

View Full Product Details