Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 2040992-2041249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH1
Entrez Gene ID
1801
Gene Name
diphthamide biosynthesis 1
Gene Aliases
DEDSSH, DPH2L, DPH2L1, OVCA1
UniGene
Hs.513856
Gene Chromosome Location
Chr.17: 2030137-2043431 on Build GRCh38
Transcripts
NM_001383.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1679368 c.1100A>G p.E367G Chr.17: 2041180-2041180 on Build GRCh38
5432104 c.1086G>A p.L362L Chr.17: 2041166-2041166 on Build GRCh38
7176071 c.1101+1G>A p.? Chr.17: 2041182-2041182 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1005920163 Chr.17: 2041029-2041029 on Build GRCh38
rs746607353 Chr.17: 2041065-2041065 on Build GRCh38
rs746700542 Chr.17: 2041191-2041191 on Build GRCh38
rs769416571 Chr.17: 2041204-2041204 on Build GRCh38
rs372258424 Chr.17: 2041053-2041053 on Build GRCh38
rs764640116 Chr.17: 2041156-2041156 on Build GRCh38
rs375928860 Chr.17: 2041110-2041110 on Build GRCh38
rs749208196 Chr.17: 2041218-2041218 on Build GRCh38
rs761845712 Chr.17: 2041091-2041091 on Build GRCh38
rs1015055805 Chr.17: 2041029-2041030 on Build GRCh38
rs575501319 Chr.17: 2041163-2041163 on Build GRCh38
rs771731558 Chr.17: 2041199-2041200 on Build GRCh38
rs543335975 Chr.17: 2041062-2041062 on Build GRCh38
rs754550744 Chr.17: 2041192-2041192 on Build GRCh38
rs979885345 Chr.17: 2041067-2041067 on Build GRCh38
rs752651024 Chr.17: 2041148-2041148 on Build GRCh38
rs979626228 Chr.17: 2041188-2041188 on Build GRCh38
rs775959248 Chr.17: 2041085-2041085 on Build GRCh38
rs766272832 Chr.17: 2041135-2041135 on Build GRCh38
rs779223248 Chr.17: 2041179-2041179 on Build GRCh38
rs780760687 Chr.17: 2041194-2041194 on Build GRCh38
rs754335301 Chr.17: 2041157-2041157 on Build GRCh38
rs545152540 Chr.17: 2041061-2041061 on Build GRCh38
rs961825902 Chr.17: 2041039-2041039 on Build GRCh38
rs751871471 Chr.17: 2041142-2041142 on Build GRCh38
rs764884949 Chr.17: 2041115-2041115 on Build GRCh38
rs747555994 Chr.17: 2041200-2041200 on Build GRCh38
rs985735275 Chr.17: 2041149-2041149 on Build GRCh38
rs767665310 Chr.17: 2041147-2041147 on Build GRCh38
rs915301730 Chr.17: 2041057-2041057 on Build GRCh38
rs773156519 Chr.17: 2041116-2041116 on Build GRCh38
rs929639649 Chr.17: 2041145-2041145 on Build GRCh38
rs911392161 Chr.17: 2041153-2041153 on Build GRCh38
rs368892850 Chr.17: 2041119-2041119 on Build GRCh38
rs770898866 Chr.17: 2041226-2041226 on Build GRCh38
rs779964465 Chr.17: 2041220-2041220 on Build GRCh38
rs901359713 Chr.17: 2041223-2041223 on Build GRCh38
rs1030011875 Chr.17: 2041060-2041060 on Build GRCh38
rs1006669505 Chr.17: 2041160-2041160 on Build GRCh38
rs373077352 Chr.17: 2041151-2041151 on Build GRCh38
rs750030837 Chr.17: 2041143-2041143 on Build GRCh38
rs773351114 Chr.17: 2041206-2041206 on Build GRCh38
rs768258224 Chr.17: 2041077-2041077 on Build GRCh38
rs946831949 Chr.17: 2041173-2041173 on Build GRCh38
rs377571866 Chr.17: 2041177-2041177 on Build GRCh38

View Full Product Details