Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129288825-129289058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMCES
Entrez Gene ID
56941
Gene Name
5-hydroxymethylcytosine (hmC) binding, ES cell-specific
Gene Aliases
C3orf37, DC12, SRAPD1
UniGene
Hs.458320
Gene Chromosome Location
Chr.3: 129278806-129305293 on Build GRCh38
Transcripts
NM_001006109.1 NM_020187.2 XM_005247636.3 XM_005247637.3 XM_017006877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5620385 c.272A>C p.Q91P Chr.3: 129288942-129288942 on Build GRCh38
1484584 c.298G>A p.D100N Chr.3: 129288968-129288968 on Build GRCh38
4945436 c.255T>C p.S85S Chr.3: 129288925-129288925 on Build GRCh38
4824856 c.197C>T p.S66F Chr.3: 129288867-129288867 on Build GRCh38
3846160 c.199G>C p.E67Q Chr.3: 129288869-129288869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752334483 Chr.3: 129289023-129289024 on Build GRCh38
rs555534959 Chr.3: 129288872-129288872 on Build GRCh38
rs755362300 Chr.3: 129289013-129289014 on Build GRCh38
rs765721619 Chr.3: 129288992-129288992 on Build GRCh38
rs761307915 Chr.3: 129288882-129288882 on Build GRCh38
rs929667184 Chr.3: 129288928-129288928 on Build GRCh38
rs1020389085 Chr.3: 129289028-129289028 on Build GRCh38
rs371706577 Chr.3: 129288986-129288986 on Build GRCh38
rs372430315 Chr.3: 129288873-129288873 on Build GRCh38
rs758851234 Chr.3: 129288906-129288906 on Build GRCh38
rs754279082 Chr.3: 129288890-129288890 on Build GRCh38
rs769720955 Chr.3: 129288963-129288963 on Build GRCh38
rs757258735 Chr.3: 129288948-129288948 on Build GRCh38
rs114856369 Chr.3: 129288987-129288987 on Build GRCh38
rs751883957 Chr.3: 129288946-129288946 on Build GRCh38
rs776149687 Chr.3: 129289016-129289016 on Build GRCh38
rs745963189 Chr.3: 129288959-129288959 on Build GRCh38
rs149403437 Chr.3: 129288878-129288878 on Build GRCh38
rs139766481 Chr.3: 129288973-129288973 on Build GRCh38
rs974256757 Chr.3: 129288886-129288886 on Build GRCh38
rs767130163 Chr.3: 129289020-129289022 on Build GRCh38
rs779979711 Chr.3: 129288972-129288972 on Build GRCh38
rs573858849 Chr.3: 129288974-129288974 on Build GRCh38
rs375669857 Chr.3: 129288884-129288884 on Build GRCh38
rs144761816 Chr.3: 129288891-129288891 on Build GRCh38
rs753065352 Chr.3: 129288897-129288897 on Build GRCh38
rs769442334 Chr.3: 129288865-129288865 on Build GRCh38
rs202031393 Chr.3: 129289021-129289021 on Build GRCh38
rs747848977 Chr.3: 129288978-129288978 on Build GRCh38
rs764599406 Chr.3: 129289024-129289024 on Build GRCh38
rs751798380 Chr.3: 129289025-129289025 on Build GRCh38
rs778078010 Chr.3: 129288936-129288936 on Build GRCh38
rs745864221 Chr.3: 129288848-129288848 on Build GRCh38
rs369330426 Chr.3: 129288950-129288950 on Build GRCh38
rs757458459 Chr.3: 129289026-129289026 on Build GRCh38
rs775312915 Chr.3: 129288866-129288866 on Build GRCh38
rs760128001 Chr.3: 129288988-129288988 on Build GRCh38

View Full Product Details