Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 174001131-174001356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf47
Entrez Gene ID
133491
Gene Name
chromosome 5 open reading frame 47
Gene Aliases
-
UniGene
Hs.131469
Gene Chromosome Location
Chr.5: 173989159-174007524 on Build GRCh38
Transcripts
NM_001144954.1 XM_011534431.1 XM_017009028.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5785155 c.512-8G>T p.? Chr.5: 174001188-174001188 on Build GRCh38
4420819 c.524C>A p.T175K Chr.5: 174001208-174001208 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs940344717 Chr.5: 174001258-174001258 on Build GRCh38
rs767962236 Chr.5: 174001248-174001248 on Build GRCh38
rs925072168 Chr.5: 174001250-174001260 on Build GRCh38
rs759796420 Chr.5: 174001247-174001247 on Build GRCh38
rs550587675 Chr.5: 174001322-174001322 on Build GRCh38
rs542797299 Chr.5: 174001205-174001205 on Build GRCh38
rs746052228 Chr.5: 174001259-174001259 on Build GRCh38
rs532390837 Chr.5: 174001288-174001288 on Build GRCh38
rs999638996 Chr.5: 174001212-174001212 on Build GRCh38
rs766787771 Chr.5: 174001216-174001216 on Build GRCh38
rs931505303 Chr.5: 174001276-174001276 on Build GRCh38
rs969686808 Chr.5: 174001228-174001228 on Build GRCh38
rs767375188 Chr.5: 174001195-174001197 on Build GRCh38
rs957933098 Chr.5: 174001221-174001221 on Build GRCh38
rs113286165 Chr.5: 174001243-174001243 on Build GRCh38
rs963174507 Chr.5: 174001292-174001292 on Build GRCh38
rs972275948 Chr.5: 174001308-174001308 on Build GRCh38
rs540143644 Chr.5: 174001260-174001260 on Build GRCh38
rs1019602463 Chr.5: 174001211-174001211 on Build GRCh38
rs575280177 Chr.5: 174001157-174001157 on Build GRCh38
rs188814530 Chr.5: 174001265-174001265 on Build GRCh38
rs1031181171 Chr.5: 174001192-174001192 on Build GRCh38
rs181186395 Chr.5: 174001318-174001318 on Build GRCh38
rs890059473 Chr.5: 174001296-174001296 on Build GRCh38
rs62378684 Chr.5: 174001225-174001225 on Build GRCh38
rs141052002 Chr.5: 174001214-174001214 on Build GRCh38
rs999522856 Chr.5: 174001325-174001325 on Build GRCh38
rs916504818 Chr.5: 174001226-174001226 on Build GRCh38

View Full Product Details