Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66285376-66285609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YIF1A
Entrez Gene ID
10897
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
Gene Aliases
54TM, FinGER7, YIF1, YIF1P
UniGene
Hs.446445
Gene Chromosome Location
Chr.11: 66284580-66289167 on Build GRCh38
Transcripts
NM_001300861.1 NM_020470.2 XM_005273720.3 XM_017017139.1 XM_017017140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7229839 c.633A>G p.K211K Chr.11: 66285389-66285389 on Build GRCh38
138005 c.554C>T p.A185V Chr.11: 66285468-66285468 on Build GRCh38
5360428 c.535delG p.V179fs*2 Chr.11: 66285487-66285487 on Build GRCh38
6306865 c.587G>A p.R196H Chr.11: 66285435-66285435 on Build GRCh38
5182202 c.586C>T p.R196C Chr.11: 66285436-66285436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745606472 Chr.11: 66285464-66285464 on Build GRCh38
rs754002854 Chr.11: 66285521-66285521 on Build GRCh38
rs148222670 Chr.11: 66285440-66285440 on Build GRCh38
rs956826334 Chr.11: 66285448-66285448 on Build GRCh38
rs748949016 Chr.11: 66285479-66285479 on Build GRCh38
rs763611495 Chr.11: 66285400-66285400 on Build GRCh38
rs774943040 Chr.11: 66285476-66285476 on Build GRCh38
rs369211299 Chr.11: 66285527-66285527 on Build GRCh38
rs768349464 Chr.11: 66285581-66285581 on Build GRCh38
rs747314467 Chr.11: 66285488-66285489 on Build GRCh38
rs757306848 Chr.11: 66285455-66285455 on Build GRCh38
rs754112960 Chr.11: 66285435-66285435 on Build GRCh38
rs145901576 Chr.11: 66285439-66285439 on Build GRCh38
rs766770020 Chr.11: 66285506-66285506 on Build GRCh38
rs550537586 Chr.11: 66285502-66285502 on Build GRCh38
rs961499601 Chr.11: 66285556-66285556 on Build GRCh38
rs201862801 Chr.11: 66285480-66285480 on Build GRCh38
rs575335774 Chr.11: 66285573-66285573 on Build GRCh38
rs748928771 Chr.11: 66285580-66285580 on Build GRCh38
rs773912505 Chr.11: 66285493-66285493 on Build GRCh38
rs779970511 Chr.11: 66285567-66285567 on Build GRCh38
rs373035564 Chr.11: 66285497-66285497 on Build GRCh38
rs755802633 Chr.11: 66285537-66285537 on Build GRCh38
rs150672448 Chr.11: 66285531-66285531 on Build GRCh38
rs762316597 Chr.11: 66285397-66285397 on Build GRCh38
rs757681308 Chr.11: 66285478-66285478 on Build GRCh38
rs755306836 Chr.11: 66285525-66285525 on Build GRCh38
rs536918442 Chr.11: 66285422-66285422 on Build GRCh38
rs377063479 Chr.11: 66285512-66285512 on Build GRCh38
rs768054964 Chr.11: 66285498-66285498 on Build GRCh38
rs768154963 Chr.11: 66285491-66285491 on Build GRCh38
rs758579591 Chr.11: 66285528-66285528 on Build GRCh38
rs141955007 Chr.11: 66285436-66285436 on Build GRCh38
rs769650237 Chr.11: 66285475-66285475 on Build GRCh38
rs756330560 Chr.11: 66285430-66285430 on Build GRCh38
rs773801488 Chr.11: 66285501-66285501 on Build GRCh38
rs750803807 Chr.11: 66285425-66285425 on Build GRCh38
rs137934642 Chr.11: 66285416-66285416 on Build GRCh38
rs556851961 Chr.11: 66285423-66285423 on Build GRCh38
rs34297861 Chr.11: 66285460-66285460 on Build GRCh38
rs570757759 Chr.11: 66285433-66285433 on Build GRCh38

View Full Product Details