Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9966419-9966646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL5A3
Entrez Gene ID
50509
Gene Name
collagen type V alpha 3
Gene Aliases
-
UniGene
Hs.235368
Gene Chromosome Location
Chr.19: 9959561-10010532 on Build GRCh38
Transcripts
NM_015719.3 XM_011528042.2 XM_017026849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4619096 c.4576G>C p.E1526Q Chr.19: 9966629-9966629 on Build GRCh38
4073690 c.4592G>A p.R1531Q Chr.19: 9966613-9966613 on Build GRCh38
3692284 c.4566G>A p.S1522S Chr.19: 9966639-9966639 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753704267 Chr.19: 9966527-9966527 on Build GRCh38
rs187502272 Chr.19: 9966469-9966469 on Build GRCh38
rs373086816 Chr.19: 9966453-9966453 on Build GRCh38
rs763206379 Chr.19: 9966494-9966494 on Build GRCh38
rs762054437 Chr.19: 9966518-9966518 on Build GRCh38
rs777776415 Chr.19: 9966448-9966448 on Build GRCh38
rs747211641 Chr.19: 9966475-9966475 on Build GRCh38
rs756176856 Chr.19: 9966519-9966519 on Build GRCh38
rs759215406 Chr.19: 9966510-9966510 on Build GRCh38
rs748118502 Chr.19: 9966439-9966439 on Build GRCh38
rs956509114 Chr.19: 9966596-9966596 on Build GRCh38
rs767139457 Chr.19: 9966517-9966517 on Build GRCh38
rs766096921 Chr.19: 9966495-9966495 on Build GRCh38
rs751202808 Chr.19: 9966503-9966503 on Build GRCh38
rs1005064639 Chr.19: 9966487-9966487 on Build GRCh38
rs201058798 Chr.19: 9966489-9966489 on Build GRCh38
rs1014280993 Chr.19: 9966490-9966490 on Build GRCh38
rs757031604 Chr.19: 9966539-9966539 on Build GRCh38
rs1024873374 Chr.19: 9966532-9966532 on Build GRCh38
rs1008797937 Chr.19: 9966603-9966603 on Build GRCh38
rs768803819 Chr.19: 9966613-9966613 on Build GRCh38
rs3745582 Chr.19: 9966482-9966482 on Build GRCh38
rs1019298477 Chr.19: 9966626-9966626 on Build GRCh38
rs980741820 Chr.19: 9966553-9966553 on Build GRCh38
rs776638360 Chr.19: 9966476-9966476 on Build GRCh38
rs528109510 Chr.19: 9966556-9966556 on Build GRCh38
rs764095590 Chr.19: 9966478-9966481 on Build GRCh38
rs1050245430 Chr.19: 9966522-9966522 on Build GRCh38
rs778717136 Chr.19: 9966549-9966549 on Build GRCh38
rs781320467 Chr.19: 9966624-9966624 on Build GRCh38
rs970652729 Chr.19: 9966547-9966547 on Build GRCh38
rs763088219 Chr.19: 9966473-9966474 on Build GRCh38
rs756048984 Chr.19: 9966441-9966441 on Build GRCh38
rs774449371 Chr.19: 9966486-9966487 on Build GRCh38
rs1000917709 Chr.19: 9966555-9966555 on Build GRCh38
rs563370281 Chr.19: 9966472-9966472 on Build GRCh38
rs747206052 Chr.19: 9966564-9966564 on Build GRCh38
rs762315981 Chr.19: 9966486-9966486 on Build GRCh38
rs904611669 Chr.19: 9966502-9966502 on Build GRCh38
rs993508361 Chr.19: 9966481-9966482 on Build GRCh38
rs752249793 Chr.19: 9966518-9966518 on Build GRCh38
rs3745583 Chr.19: 9966523-9966523 on Build GRCh38
rs937580773 Chr.19: 9966477-9966477 on Build GRCh38
rs765731246 Chr.19: 9966488-9966488 on Build GRCh38
rs920958352 Chr.19: 9966450-9966450 on Build GRCh38
rs972334856 Chr.19: 9966524-9966524 on Build GRCh38
rs987954158 Chr.19: 9966612-9966612 on Build GRCh38
rs868593655 Chr.19: 9966548-9966548 on Build GRCh38

View Full Product Details