Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 77282185-77282436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK5
Entrez Gene ID
26289
Gene Name
adenylate kinase 5
Gene Aliases
AK6
UniGene
Hs.559718
Gene Chromosome Location
Chr.1: 77281908-77559969 on Build GRCh38
Transcripts
NM_174858.2 XM_005270739.4 XM_017001010.1 XM_017001012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182336 c.38G>T p.R13M Chr.1: 77282351-77282351 on Build GRCh38
3689839 c.5A>T p.N2I Chr.1: 77282318-77282318 on Build GRCh38
426699 c.58G>C p.E20Q Chr.1: 77282371-77282371 on Build GRCh38
6597352 c.12C>T p.N4N Chr.1: 77282325-77282325 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1048182487 Chr.1: 77282393-77282393 on Build GRCh38
rs780630971 Chr.1: 77282399-77282399 on Build GRCh38
rs573383156 Chr.1: 77282319-77282319 on Build GRCh38
rs987107368 Chr.1: 77282217-77282217 on Build GRCh38
rs896545392 Chr.1: 77282315-77282315 on Build GRCh38
rs1056323489 Chr.1: 77282299-77282299 on Build GRCh38
rs61784593 Chr.1: 77282394-77282394 on Build GRCh38
rs764955919 Chr.1: 77282264-77282264 on Build GRCh38
rs557054752 Chr.1: 77282203-77282203 on Build GRCh38
rs752283975 Chr.1: 77282268-77282268 on Build GRCh38
rs868102635 Chr.1: 77282345-77282345 on Build GRCh38
rs114518551 Chr.1: 77282218-77282218 on Build GRCh38
rs926143895 Chr.1: 77282294-77282294 on Build GRCh38
rs915904952 Chr.1: 77282363-77282363 on Build GRCh38
rs577019614 Chr.1: 77282383-77282383 on Build GRCh38
rs575287320 Chr.1: 77282211-77282211 on Build GRCh38
rs769046602 Chr.1: 77282402-77282402 on Build GRCh38
rs967624355 Chr.1: 77282226-77282226 on Build GRCh38
rs1056573051 Chr.1: 77282314-77282314 on Build GRCh38
rs1043917983 Chr.1: 77282261-77282261 on Build GRCh38
rs749507463 Chr.1: 77282401-77282401 on Build GRCh38
rs949996867 Chr.1: 77282391-77282391 on Build GRCh38
rs758117727 Chr.1: 77282296-77282296 on Build GRCh38
rs116049705 Chr.1: 77282297-77282297 on Build GRCh38
rs774542727 Chr.1: 77282413-77282413 on Build GRCh38
rs946969412 Chr.1: 77282230-77282230 on Build GRCh38
rs781687353 Chr.1: 77282321-77282321 on Build GRCh38
rs746574186 Chr.1: 77282309-77282309 on Build GRCh38

View Full Product Details