Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44667370-44667617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEIOC
Entrez Gene ID
284071
Gene Name
meiosis specific with coiled-coil domain
Gene Aliases
C17orf104
UniGene
Hs.349758
Gene Chromosome Location
Chr.17: 44656394-44677086 on Build GRCh38
Transcripts
NM_001145080.2 XM_005257236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
980008 c.1625C>T p.A542V Chr.17: 44667536-44667536 on Build GRCh38
1198221 c.1507C>A p.H503N Chr.17: 44667418-44667418 on Build GRCh38
1283740 c.1469C>T p.P490L Chr.17: 44667380-44667380 on Build GRCh38
3191338 c.1603T>G p.Y535D Chr.17: 44667514-44667514 on Build GRCh38
187768 c.1474C>T p.R492* Chr.17: 44667385-44667385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768512026 Chr.17: 44667524-44667524 on Build GRCh38
rs369478803 Chr.17: 44667483-44667483 on Build GRCh38
rs764520453 Chr.17: 44667571-44667571 on Build GRCh38
rs376503425 Chr.17: 44667528-44667528 on Build GRCh38
rs762468165 Chr.17: 44667588-44667588 on Build GRCh38
rs776848317 Chr.17: 44667492-44667492 on Build GRCh38
rs1003002405 Chr.17: 44667566-44667566 on Build GRCh38
rs764314659 Chr.17: 44667429-44667429 on Build GRCh38
rs752124964 Chr.17: 44667430-44667430 on Build GRCh38
rs749424821 Chr.17: 44667512-44667512 on Build GRCh38
rs1038206041 Chr.17: 44667471-44667471 on Build GRCh38
rs757770823 Chr.17: 44667448-44667448 on Build GRCh38
rs772762258 Chr.17: 44667549-44667549 on Build GRCh38
rs750569354 Chr.17: 44667592-44667592 on Build GRCh38
rs377221459 Chr.17: 44667572-44667572 on Build GRCh38
rs540773024 Chr.17: 44667500-44667500 on Build GRCh38
rs372682730 Chr.17: 44667462-44667462 on Build GRCh38
rs368948349 Chr.17: 44667590-44667590 on Build GRCh38
rs772709127 Chr.17: 44667409-44667409 on Build GRCh38
rs780288556 Chr.17: 44667504-44667504 on Build GRCh38
rs751963734 Chr.17: 44667587-44667587 on Build GRCh38
rs765769398 Chr.17: 44667563-44667563 on Build GRCh38
rs759050083 Chr.17: 44667565-44667565 on Build GRCh38
rs758825117 Chr.17: 44667427-44667427 on Build GRCh38
rs971453983 Chr.17: 44667516-44667516 on Build GRCh38
rs138599725 Chr.17: 44667551-44667551 on Build GRCh38
rs920584357 Chr.17: 44667476-44667476 on Build GRCh38
rs771988635 Chr.17: 44667545-44667545 on Build GRCh38
rs776366790 Chr.17: 44667564-44667564 on Build GRCh38
rs753045227 Chr.17: 44667425-44667425 on Build GRCh38
rs760377974 Chr.17: 44667419-44667419 on Build GRCh38
rs766033557 Chr.17: 44667424-44667424 on Build GRCh38
rs372925968 Chr.17: 44667539-44667539 on Build GRCh38
rs749749473 Chr.17: 44667404-44667409 on Build GRCh38

View Full Product Details