Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 21760339-21760612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH12
Entrez Gene ID
1010
Gene Name
cadherin 12
Gene Aliases
CDHB
UniGene
Hs.113684
Gene Chromosome Location
Chr.5: 21750191-22853622 on Build GRCh38
Transcripts
NM_001317227.1 NM_001317228.1 NM_004061.4 XM_011513927.2 XM_017008920.1 XM_017008921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6447176 c.1621C>T p.R541C Chr.5: 21760570-21760570 on Build GRCh38
368890 c.1624_1625GA>TT p.D542>? Chr.5: 21760566-21760567 on Build GRCh38
3725706 c.1607C>A p.P536Q Chr.5: 21760584-21760584 on Build GRCh38
737316 c.1604A>T p.K535I Chr.5: 21760587-21760587 on Build GRCh38
5818397 c.1599T>C p.A533A Chr.5: 21760592-21760592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896664554 Chr.5: 21760428-21760428 on Build GRCh38
rs543432386 Chr.5: 21760379-21760379 on Build GRCh38
rs1043820147 Chr.5: 21760377-21760377 on Build GRCh38
rs778998092 Chr.5: 21760529-21760529 on Build GRCh38
rs952338016 Chr.5: 21760485-21760485 on Build GRCh38
rs904847371 Chr.5: 21760382-21760382 on Build GRCh38
rs533294662 Chr.5: 21760587-21760587 on Build GRCh38
rs758334069 Chr.5: 21760533-21760533 on Build GRCh38
rs375131865 Chr.5: 21760512-21760512 on Build GRCh38
rs1034734747 Chr.5: 21760418-21760418 on Build GRCh38
rs200531657 Chr.5: 21760546-21760546 on Build GRCh38
rs369955895 Chr.5: 21760570-21760570 on Build GRCh38
rs1026427922 Chr.5: 21760484-21760484 on Build GRCh38
rs369407051 Chr.5: 21760510-21760510 on Build GRCh38
rs559400881 Chr.5: 21760505-21760505 on Build GRCh38
rs768854360 Chr.5: 21760522-21760524 on Build GRCh38
rs112912165 Chr.5: 21760388-21760388 on Build GRCh38
rs573960884 Chr.5: 21760448-21760448 on Build GRCh38
rs140423691 Chr.5: 21760573-21760573 on Build GRCh38
rs956098353 Chr.5: 21760535-21760535 on Build GRCh38
rs763341567 Chr.5: 21760527-21760527 on Build GRCh38
rs946600199 Chr.5: 21760368-21760368 on Build GRCh38
rs2303748 Chr.5: 21760493-21760493 on Build GRCh38
rs993597510 Chr.5: 21760445-21760445 on Build GRCh38
rs375154168 Chr.5: 21760547-21760547 on Build GRCh38
rs200808789 Chr.5: 21760569-21760569 on Build GRCh38
rs369535033 Chr.5: 21760539-21760539 on Build GRCh38
rs750300677 Chr.5: 21760532-21760532 on Build GRCh38

View Full Product Details