Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134738504-134738769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMLG
Entrez Gene ID
819
Gene Name
calcium modulating ligand
Gene Aliases
CAML, GET2
UniGene
Hs.529846
Gene Chromosome Location
Chr.5: 134737927-134752160 on Build GRCh38
Transcripts
NM_001745.3 XM_011543653.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7359359 c.31G>C p.G11R Chr.5: 134738651-134738651 on Build GRCh38
1433190 c.71C>T p.A24V Chr.5: 134738691-134738691 on Build GRCh38
388672 c.5A>G p.E2G Chr.5: 134738625-134738625 on Build GRCh38
1310555 c.87G>A p.A29A Chr.5: 134738707-134738707 on Build GRCh38
6295052 c.93G>T p.L31L Chr.5: 134738713-134738713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750285729 Chr.5: 134738728-134738728 on Build GRCh38
rs181306113 Chr.5: 134738617-134738617 on Build GRCh38
rs774232800 Chr.5: 134738717-134738717 on Build GRCh38
rs1037934098 Chr.5: 134738524-134738524 on Build GRCh38
rs966908374 Chr.5: 134738576-134738576 on Build GRCh38
rs550990078 Chr.5: 134738638-134738638 on Build GRCh38
rs899030332 Chr.5: 134738526-134738526 on Build GRCh38
rs762873568 Chr.5: 134738662-134738662 on Build GRCh38
rs771353868 Chr.5: 134738647-134738647 on Build GRCh38
rs778378306 Chr.5: 134738697-134738697 on Build GRCh38
rs1011191683 Chr.5: 134738561-134738561 on Build GRCh38
rs75279757 Chr.5: 134738667-134738667 on Build GRCh38
rs546356304 Chr.5: 134738629-134738629 on Build GRCh38
rs563884241 Chr.5: 134738732-134738732 on Build GRCh38
rs368323992 Chr.5: 134738715-134738715 on Build GRCh38
rs895385442 Chr.5: 134738557-134738557 on Build GRCh38
rs767156487 Chr.5: 134738727-134738727 on Build GRCh38
rs761174350 Chr.5: 134738641-134738641 on Build GRCh38
rs775518646 Chr.5: 134738709-134738709 on Build GRCh38
rs757841175 Chr.5: 134738741-134738741 on Build GRCh38
rs765392326 Chr.5: 134738582-134738582 on Build GRCh38
rs1031440293 Chr.5: 134738601-134738601 on Build GRCh38
rs1053798697 Chr.5: 134738531-134738531 on Build GRCh38
rs200925040 Chr.5: 134738688-134738688 on Build GRCh38
rs936940629 Chr.5: 134738685-134738685 on Build GRCh38
rs746065550 Chr.5: 134738706-134738706 on Build GRCh38
rs753936018 Chr.5: 134738574-134738574 on Build GRCh38
rs559785805 Chr.5: 134738589-134738589 on Build GRCh38
rs370682989 Chr.5: 134738700-134738700 on Build GRCh38
rs143920630 Chr.5: 134738702-134738702 on Build GRCh38
rs28384481 Chr.5: 134738538-134738538 on Build GRCh38
rs901251038 Chr.5: 134738544-134738544 on Build GRCh38
rs766573194 Chr.5: 134738570-134738570 on Build GRCh38
rs368615673 Chr.5: 134738708-134738708 on Build GRCh38
rs372065125 Chr.5: 134738551-134738551 on Build GRCh38
rs746285572 Chr.5: 134738750-134738750 on Build GRCh38
rs111282208 Chr.5: 134738580-134738580 on Build GRCh38
rs772673231 Chr.5: 134738696-134738696 on Build GRCh38
rs777531400 Chr.5: 134738592-134738592 on Build GRCh38
rs985080818 Chr.5: 134738558-134738558 on Build GRCh38
rs780939456 Chr.5: 134738553-134738553 on Build GRCh38
rs531402508 Chr.5: 134738733-134738733 on Build GRCh38
rs761813309 Chr.5: 134738723-134738723 on Build GRCh38
rs866155126 Chr.5: 134738690-134738690 on Build GRCh38
rs1033665130 Chr.5: 134738556-134738556 on Build GRCh38
rs780756030 Chr.5: 134738603-134738603 on Build GRCh38
rs990599789 Chr.5: 134738606-134738606 on Build GRCh38
rs370920966 Chr.5: 134738621-134738621 on Build GRCh38
rs756995409 Chr.5: 134738666-134738666 on Build GRCh38
rs772759398 Chr.5: 134738639-134738639 on Build GRCh38
rs751332405 Chr.5: 134738665-134738665 on Build GRCh38
rs141336392 Chr.5: 134738653-134738653 on Build GRCh38
rs779413552 Chr.5: 134738684-134738684 on Build GRCh38
rs781406751 Chr.5: 134738742-134738742 on Build GRCh38
rs777067740 Chr.5: 134738705-134738705 on Build GRCh38
rs987114990 Chr.5: 134738626-134738626 on Build GRCh38
rs201737362 Chr.5: 134738731-134738731 on Build GRCh38
rs374901242 Chr.5: 134738740-134738740 on Build GRCh38
rs754561770 Chr.5: 134738536-134738536 on Build GRCh38
rs763069452 Chr.5: 134738711-134738711 on Build GRCh38
rs781071324 Chr.5: 134738676-134738676 on Build GRCh38
rs943270359 Chr.5: 134738644-134738644 on Build GRCh38
rs776679756 Chr.5: 134738650-134738650 on Build GRCh38
rs751422675 Chr.5: 134738597-134738597 on Build GRCh38
rs745555514 Chr.5: 134738607-134738607 on Build GRCh38
rs773704477 Chr.5: 134738640-134738640 on Build GRCh38
rs560294130 Chr.5: 134738542-134738542 on Build GRCh38
rs752511826 Chr.5: 134738585-134738585 on Build GRCh38
rs755800357 Chr.5: 134738679-134738679 on Build GRCh38
rs929347200 Chr.5: 134738539-134738539 on Build GRCh38
rs376579627 Chr.5: 134738669-134738669 on Build GRCh38
rs759488048 Chr.5: 134738577-134738577 on Build GRCh38
rs375153038 Chr.5: 134738651-134738651 on Build GRCh38
rs771138673 Chr.5: 134738704-134738704 on Build GRCh38
rs749975874 Chr.5: 134738677-134738677 on Build GRCh38
rs144459434 Chr.5: 134738583-134738584 on Build GRCh38
rs576803974 Chr.5: 134738529-134738529 on Build GRCh38
rs752834014 Chr.5: 134738658-134738658 on Build GRCh38

View Full Product Details