Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35340942-35341205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD22
Entrez Gene ID
933
Gene Name
CD22 molecule
Gene Aliases
SIGLEC-2, SIGLEC2
UniGene
Hs.579691
Gene Chromosome Location
Chr.19: 35329166-35347361 on Build GRCh38
Transcripts
NM_001185099.1 NM_001185100.1 NM_001185101.1 NM_001278417.1 NM_001771.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4808411 c.1507T>C p.Y503H Chr.19: 35341138-35341138 on Build GRCh38
187873 c.1422C>T p.N474N Chr.19: 35341053-35341053 on Build GRCh38
7289858 c.1366T>C p.W456R Chr.19: 35340997-35340997 on Build GRCh38
6249694 c.1403G>T p.G468V Chr.19: 35341034-35341034 on Build GRCh38
7268044 c.1357C>T p.R453W Chr.19: 35340988-35340988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759692965 Chr.19: 35340980-35340980 on Build GRCh38
rs761096870 Chr.19: 35341100-35341100 on Build GRCh38
rs141116950 Chr.19: 35340975-35340975 on Build GRCh38
rs766149151 Chr.19: 35341106-35341106 on Build GRCh38
rs766954871 Chr.19: 35341055-35341055 on Build GRCh38
rs764742378 Chr.19: 35341117-35341117 on Build GRCh38
rs201759195 Chr.19: 35341058-35341058 on Build GRCh38
rs978270422 Chr.19: 35341017-35341017 on Build GRCh38
rs777598834 Chr.19: 35341054-35341054 on Build GRCh38
rs771859129 Chr.19: 35340978-35340978 on Build GRCh38
rs768749232 Chr.19: 35341184-35341184 on Build GRCh38
rs750804257 Chr.19: 35341021-35341021 on Build GRCh38
rs748051201 Chr.19: 35341034-35341034 on Build GRCh38
rs199643177 Chr.19: 35341053-35341053 on Build GRCh38
rs144878916 Chr.19: 35341028-35341028 on Build GRCh38
rs991082304 Chr.19: 35341036-35341036 on Build GRCh38
rs200102428 Chr.19: 35341098-35341098 on Build GRCh38
rs746831747 Chr.19: 35341169-35341169 on Build GRCh38
rs754450480 Chr.19: 35341112-35341112 on Build GRCh38
rs25677 Chr.19: 35341083-35341083 on Build GRCh38
rs768718326 Chr.19: 35341079-35341079 on Build GRCh38
rs187073369 Chr.19: 35340987-35340987 on Build GRCh38
rs370742313 Chr.19: 35341012-35341012 on Build GRCh38
rs769987261 Chr.19: 35340985-35340985 on Build GRCh38
rs762440332 Chr.19: 35341088-35341088 on Build GRCh38
rs758394183 Chr.19: 35341145-35341145 on Build GRCh38
rs775643440 Chr.19: 35341077-35341077 on Build GRCh38
rs755899640 Chr.19: 35341027-35341027 on Build GRCh38
rs140331196 Chr.19: 35341084-35341084 on Build GRCh38
rs371948951 Chr.19: 35341179-35341179 on Build GRCh38
rs369338988 Chr.19: 35341162-35341162 on Build GRCh38
rs374081081 Chr.19: 35341029-35341029 on Build GRCh38
rs767936787 Chr.19: 35341018-35341018 on Build GRCh38
rs564242435 Chr.19: 35341033-35341033 on Build GRCh38
rs370636164 Chr.19: 35341060-35341060 on Build GRCh38
rs749363464 Chr.19: 35341078-35341078 on Build GRCh38
rs776839232 Chr.19: 35340979-35340979 on Build GRCh38
rs777654032 Chr.19: 35341151-35341151 on Build GRCh38
rs369908101 Chr.19: 35340989-35340989 on Build GRCh38
rs780264204 Chr.19: 35341180-35341180 on Build GRCh38
rs768135426 Chr.19: 35341097-35341097 on Build GRCh38
rs763068399 Chr.19: 35340988-35340988 on Build GRCh38
rs752070697 Chr.19: 35341011-35341011 on Build GRCh38
rs376922176 Chr.19: 35341107-35341107 on Build GRCh38
rs760563783 Chr.19: 35341123-35341123 on Build GRCh38
rs752131543 Chr.19: 35341137-35341137 on Build GRCh38
rs778886530 Chr.19: 35340974-35340974 on Build GRCh38

View Full Product Details