Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 6876044-6876283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP28
Entrez Gene ID
79822
Gene Name
Rho GTPase activating protein 28
Gene Aliases
-
UniGene
Hs.183114
Gene Chromosome Location
Chr.18: 6729718-6915716 on Build GRCh38
Transcripts
NM_001010000.2 XM_005258144.1 XM_005258145.1 XM_005258146.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
278949 c.1215T>G p.F405L Chr.18: 6876133-6876133 on Build GRCh38
380432 c.1253G>T p.G418V Chr.18: 6876171-6876171 on Build GRCh38
417801 c.742G>C p.E248Q Chr.18: 6876137-6876137 on Build GRCh38
254537 c.1249G>T p.E417* Chr.18: 6876167-6876167 on Build GRCh38
1245353 c.747A>C p.K249N Chr.18: 6876142-6876142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149236554 Chr.18: 6876191-6876191 on Build GRCh38
rs771523079 Chr.18: 6876214-6876214 on Build GRCh38
rs758359951 Chr.18: 6876133-6876133 on Build GRCh38
rs781727091 Chr.18: 6876179-6876179 on Build GRCh38
rs889678792 Chr.18: 6876263-6876263 on Build GRCh38
rs565152011 Chr.18: 6876109-6876109 on Build GRCh38
rs760665719 Chr.18: 6876258-6876258 on Build GRCh38
rs757813579 Chr.18: 6876173-6876173 on Build GRCh38
rs925950816 Chr.18: 6876123-6876123 on Build GRCh38
rs575478884 Chr.18: 6876127-6876127 on Build GRCh38
rs772688620 Chr.18: 6876238-6876238 on Build GRCh38
rs752833545 Chr.18: 6876131-6876131 on Build GRCh38
rs1050960455 Chr.18: 6876248-6876248 on Build GRCh38
rs755262540 Chr.18: 6876118-6876118 on Build GRCh38
rs371304813 Chr.18: 6876149-6876149 on Build GRCh38
rs773920069 Chr.18: 6876089-6876089 on Build GRCh38
rs761173813 Chr.18: 6876091-6876091 on Build GRCh38
rs760575851 Chr.18: 6876097-6876097 on Build GRCh38
rs529726145 Chr.18: 6876262-6876262 on Build GRCh38
rs750325331 Chr.18: 6876095-6876095 on Build GRCh38
rs138868164 Chr.18: 6876203-6876203 on Build GRCh38
rs374208184 Chr.18: 6876213-6876213 on Build GRCh38
rs778375590 Chr.18: 6876143-6876143 on Build GRCh38
rs766212247 Chr.18: 6876108-6876108 on Build GRCh38
rs767516056 Chr.18: 6876093-6876093 on Build GRCh38
rs746170150 Chr.18: 6876180-6876180 on Build GRCh38

View Full Product Details