Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36170953-36171182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCIN
Entrez Gene ID
881
Gene Name
calicin
Gene Aliases
BTBD20, KBTBD14
UniGene
Hs.115460
Gene Chromosome Location
Chr.9: 36169392-36171334 on Build GRCh38
Transcripts
NM_005893.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3745998 c.1469T>C p.I490T Chr.9: 36170971-36170971 on Build GRCh38
5695244 c.1538T>C p.L513S Chr.9: 36171040-36171040 on Build GRCh38
4608057 c.1559G>T p.C520F Chr.9: 36171061-36171061 on Build GRCh38
6044949 c.1573A>G p.S525G Chr.9: 36171075-36171075 on Build GRCh38
3907316 c.1532G>A p.C511Y Chr.9: 36171034-36171034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372164853 Chr.9: 36171068-36171068 on Build GRCh38
rs569979444 Chr.9: 36171073-36171073 on Build GRCh38
rs534048769 Chr.9: 36171142-36171142 on Build GRCh38
rs376552213 Chr.9: 36171134-36171134 on Build GRCh38
rs774946675 Chr.9: 36171130-36171130 on Build GRCh38
rs374805321 Chr.9: 36171089-36171089 on Build GRCh38
rs761796443 Chr.9: 36171152-36171152 on Build GRCh38
rs750457920 Chr.9: 36171055-36171055 on Build GRCh38
rs764030343 Chr.9: 36171032-36171032 on Build GRCh38
rs773964161 Chr.9: 36171005-36171005 on Build GRCh38
rs781773322 Chr.9: 36171076-36171076 on Build GRCh38
rs370958730 Chr.9: 36170975-36170975 on Build GRCh38
rs765031801 Chr.9: 36171076-36171091 on Build GRCh38
rs765350319 Chr.9: 36171044-36171044 on Build GRCh38
rs983883772 Chr.9: 36171067-36171067 on Build GRCh38
rs61735202 Chr.9: 36171095-36171095 on Build GRCh38
rs374631870 Chr.9: 36170990-36170990 on Build GRCh38
rs369529709 Chr.9: 36171108-36171108 on Build GRCh38
rs112488545 Chr.9: 36171110-36171110 on Build GRCh38
rs760319760 Chr.9: 36171031-36171031 on Build GRCh38
rs755349582 Chr.9: 36171074-36171074 on Build GRCh38
rs758715079 Chr.9: 36171057-36171057 on Build GRCh38
rs761911001 Chr.9: 36171042-36171042 on Build GRCh38
rs199963031 Chr.9: 36171001-36171001 on Build GRCh38
rs769719968 Chr.9: 36171159-36171159 on Build GRCh38
rs1002663010 Chr.9: 36171069-36171069 on Build GRCh38
rs768400236 Chr.9: 36171138-36171138 on Build GRCh38
rs532897023 Chr.9: 36171002-36171002 on Build GRCh38
rs200945259 Chr.9: 36171113-36171113 on Build GRCh38
rs145792420 Chr.9: 36171029-36171029 on Build GRCh38
rs202080482 Chr.9: 36171028-36171028 on Build GRCh38

View Full Product Details