Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 44629450-44629681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YIPF7
Entrez Gene ID
285525
Gene Name
Yip1 domain family member 7
Gene Aliases
FinGER9
UniGene
Hs.596000
Gene Chromosome Location
Chr.4: 44619578-44656868 on Build GRCh38
Transcripts
NM_182592.2 XM_011513679.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5030725 c.407_408insA p.M137fs*11 Chr.4: 44629493-44629494 on Build GRCh38
4454100 c.404A>G p.N135S Chr.4: 44629497-44629497 on Build GRCh38
2848279 c.384A>T p.Q128H Chr.4: 44629517-44629517 on Build GRCh38
5663806 c.449T>G p.L150R Chr.4: 44629452-44629452 on Build GRCh38
5605364 c.424G>A p.G142S Chr.4: 44629477-44629477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754319719 Chr.4: 44629592-44629592 on Build GRCh38
rs775290678 Chr.4: 44629526-44629526 on Build GRCh38
rs763768031 Chr.4: 44629513-44629513 on Build GRCh38
rs575244981 Chr.4: 44629648-44629648 on Build GRCh38
rs767208136 Chr.4: 44629577-44629577 on Build GRCh38
rs140111997 Chr.4: 44629585-44629585 on Build GRCh38
rs557439975 Chr.4: 44629593-44629593 on Build GRCh38
rs758710102 Chr.4: 44629559-44629560 on Build GRCh38
rs371366008 Chr.4: 44629508-44629508 on Build GRCh38
rs571016096 Chr.4: 44629575-44629575 on Build GRCh38
rs932356178 Chr.4: 44629643-44629643 on Build GRCh38
rs959019714 Chr.4: 44629492-44629492 on Build GRCh38
rs760315752 Chr.4: 44629512-44629513 on Build GRCh38
rs748307835 Chr.4: 44629494-44629494 on Build GRCh38
rs1017904368 Chr.4: 44629552-44629552 on Build GRCh38
rs779143117 Chr.4: 44629481-44629481 on Build GRCh38
rs373476155 Chr.4: 44629581-44629581 on Build GRCh38
rs368551910 Chr.4: 44629505-44629505 on Build GRCh38
rs368868075 Chr.4: 44629576-44629576 on Build GRCh38
rs6813809 Chr.4: 44629570-44629570 on Build GRCh38
rs766899742 Chr.4: 44629576-44629578 on Build GRCh38
rs914541462 Chr.4: 44629524-44629524 on Build GRCh38
rs750797715 Chr.4: 44629552-44629553 on Build GRCh38
rs533892739 Chr.4: 44629523-44629523 on Build GRCh38
rs867875542 Chr.4: 44629549-44629549 on Build GRCh38
rs763782279 Chr.4: 44629536-44629536 on Build GRCh38
rs749788009 Chr.4: 44629618-44629618 on Build GRCh38
rs762750769 Chr.4: 44629528-44629528 on Build GRCh38
rs749887683 Chr.4: 44629578-44629578 on Build GRCh38
rs769810318 Chr.4: 44629514-44629514 on Build GRCh38

View Full Product Details