Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3492592-3492849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK7
Entrez Gene ID
285489
Gene Name
docking protein 7
Gene Aliases
C4orf25, CMS10, CMS1B
UniGene
Hs.122110
Gene Chromosome Location
Chr.4: 3463306-3501476 on Build GRCh38
Transcripts
NM_001164673.1 NM_001256896.1 NM_001301071.1 NM_173660.4 XM_011513435.2 XM_011513437.2 XM_017008038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1594530 c.810_811delCA p.H272fs*28 Chr.4: 3492796-3492797 on Build GRCh38
373147 c.818T>A p.L273* Chr.4: 3492804-3492804 on Build GRCh38
1054936 c.810_811delCA p.H272fs*28 Chr.4: 3492796-3492797 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753417285 Chr.4: 3492715-3492715 on Build GRCh38
rs904083238 Chr.4: 3492702-3492702 on Build GRCh38
rs763936252 Chr.4: 3492783-3492783 on Build GRCh38
rs756253716 Chr.4: 3492797-3492797 on Build GRCh38
rs778354509 Chr.4: 3492723-3492724 on Build GRCh38
rs144269117 Chr.4: 3492825-3492825 on Build GRCh38
rs777717708 Chr.4: 3492720-3492720 on Build GRCh38
rs139573161 Chr.4: 3492824-3492824 on Build GRCh38
rs150549589 Chr.4: 3492817-3492817 on Build GRCh38
rs138794794 Chr.4: 3492794-3492794 on Build GRCh38
rs560074829 Chr.4: 3492640-3492640 on Build GRCh38
rs757270887 Chr.4: 3492724-3492724 on Build GRCh38
rs555233966 Chr.4: 3492619-3492619 on Build GRCh38
rs921234407 Chr.4: 3492775-3492775 on Build GRCh38
rs192979908 Chr.4: 3492722-3492722 on Build GRCh38
rs749706359 Chr.4: 3492741-3492741 on Build GRCh38
rs141147662 Chr.4: 3492832-3492832 on Build GRCh38
rs73195197 Chr.4: 3492708-3492708 on Build GRCh38
rs971287689 Chr.4: 3492729-3492729 on Build GRCh38
rs142821143 Chr.4: 3492767-3492767 on Build GRCh38
rs3135163 Chr.4: 3492822-3492822 on Build GRCh38
rs752449499 Chr.4: 3492780-3492780 on Build GRCh38
rs115614731 Chr.4: 3492802-3492802 on Build GRCh38
rs62272676 Chr.4: 3492692-3492692 on Build GRCh38
rs748569461 Chr.4: 3492812-3492812 on Build GRCh38
rs750076866 Chr.4: 3492725-3492726 on Build GRCh38
rs757928721 Chr.4: 3492745-3492746 on Build GRCh38
rs780520331 Chr.4: 3492739-3492739 on Build GRCh38
rs779759950 Chr.4: 3492748-3492749 on Build GRCh38
rs1054593867 Chr.4: 3492648-3492648 on Build GRCh38
rs778227501 Chr.4: 3492796-3492797 on Build GRCh38
rs759117081 Chr.4: 3492815-3492815 on Build GRCh38
rs751582204 Chr.4: 3492723-3492723 on Build GRCh38
rs13146874 Chr.4: 3492623-3492623 on Build GRCh38
rs762464191 Chr.4: 3492760-3492760 on Build GRCh38
rs761609438 Chr.4: 3492827-3492827 on Build GRCh38
rs376190589 Chr.4: 3492750-3492750 on Build GRCh38
rs141732576 Chr.4: 3492808-3492808 on Build GRCh38
rs3123331 Chr.4: 3492819-3492819 on Build GRCh38
rs762740241 Chr.4: 3492781-3492781 on Build GRCh38
rs13115832 Chr.4: 3492617-3492617 on Build GRCh38
rs781401847 Chr.4: 3492726-3492726 on Build GRCh38
rs1042925470 Chr.4: 3492803-3492803 on Build GRCh38
rs891235676 Chr.4: 3492805-3492805 on Build GRCh38
rs886038745 Chr.4: 3492727-3492727 on Build GRCh38
rs894235000 Chr.4: 3492712-3492712 on Build GRCh38
rs912693449 Chr.4: 3492661-3492661 on Build GRCh38
rs6830812 Chr.4: 3492703-3492703 on Build GRCh38
rs921368686 Chr.4: 3492643-3492643 on Build GRCh38
rs936295024 Chr.4: 3492772-3492772 on Build GRCh38
rs781113489 Chr.4: 3492790-3492790 on Build GRCh38
rs936166535 Chr.4: 3492641-3492649 on Build GRCh38
rs764962641 Chr.4: 3492779-3492779 on Build GRCh38
rs144592743 Chr.4: 3492798-3492798 on Build GRCh38
rs866732188 Chr.4: 3492787-3492787 on Build GRCh38
rs751494151 Chr.4: 3492789-3492789 on Build GRCh38
rs775008753 Chr.4: 3492752-3492752 on Build GRCh38
rs111522010 Chr.4: 3492655-3492655 on Build GRCh38
rs141979124 Chr.4: 3492795-3492795 on Build GRCh38
rs772682276 Chr.4: 3492814-3492814 on Build GRCh38
rs765994305 Chr.4: 3492713-3492713 on Build GRCh38
rs16844460 Chr.4: 3492768-3492768 on Build GRCh38
rs755315038 Chr.4: 3492652-3492652 on Build GRCh38
rs770163440 Chr.4: 3492757-3492758 on Build GRCh38
rs545862751 Chr.4: 3492686-3492686 on Build GRCh38
rs370102558 Chr.4: 3492809-3492809 on Build GRCh38
rs34230391 Chr.4: 3492723-3492723 on Build GRCh38
rs764908663 Chr.4: 3492716-3492716 on Build GRCh38
rs2699415 Chr.4: 3492628-3492628 on Build GRCh38
rs748418500 Chr.4: 3492749-3492752 on Build GRCh38
rs372189971 Chr.4: 3492630-3492630 on Build GRCh38
rs368478410 Chr.4: 3492717-3492717 on Build GRCh38
rs746049063 Chr.4: 3492736-3492736 on Build GRCh38
rs532097176 Chr.4: 3492678-3492679 on Build GRCh38

View Full Product Details