Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66282010-66282283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNIH2
Entrez Gene ID
254263
Gene Name
cornichon family AMPA receptor auxiliary protein 2
Gene Aliases
CNIH-2, Cnil
UniGene
Hs.437072
Gene Chromosome Location
Chr.11: 66278201-66284214 on Build GRCh38
Transcripts
NM_182553.2 NR_073078.1 NR_073079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
237212 c.90C>T p.A30A Chr.11: 66282267-66282267 on Build GRCh38
3986370 c.87A>G p.I29M Chr.11: 66282264-66282264 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749971974 Chr.11: 66282241-66282241 on Build GRCh38
rs779416405 Chr.11: 66282246-66282246 on Build GRCh38
rs973465699 Chr.11: 66282130-66282130 on Build GRCh38
rs760957918 Chr.11: 66282211-66282211 on Build GRCh38
rs999939267 Chr.11: 66282247-66282247 on Build GRCh38
rs1038222672 Chr.11: 66282119-66282119 on Build GRCh38
rs369497283 Chr.11: 66282215-66282215 on Build GRCh38
rs1050767836 Chr.11: 66282152-66282152 on Build GRCh38
rs764061061 Chr.11: 66282219-66282219 on Build GRCh38
rs534389711 Chr.11: 66282118-66282118 on Build GRCh38
rs574076963 Chr.11: 66282168-66282168 on Build GRCh38
rs991183077 Chr.11: 66282053-66282053 on Build GRCh38
rs939270462 Chr.11: 66282037-66282037 on Build GRCh38
rs919250406 Chr.11: 66282162-66282162 on Build GRCh38
rs372970142 Chr.11: 66282240-66282240 on Build GRCh38
rs755858521 Chr.11: 66282245-66282245 on Build GRCh38
rs1007143211 Chr.11: 66282169-66282169 on Build GRCh38
rs751532644 Chr.11: 66282225-66282225 on Build GRCh38
rs929359463 Chr.11: 66282191-66282191 on Build GRCh38
rs916503523 Chr.11: 66282054-66282054 on Build GRCh38
rs112038128 Chr.11: 66282133-66282133 on Build GRCh38
rs200351390 Chr.11: 66282217-66282217 on Build GRCh38
rs868009223 Chr.11: 66282088-66282088 on Build GRCh38
rs369384568 Chr.11: 66282202-66282202 on Build GRCh38
rs756950072 Chr.11: 66282237-66282237 on Build GRCh38
rs116598987 Chr.11: 66282212-66282212 on Build GRCh38
rs374842460 Chr.11: 66282132-66282132 on Build GRCh38
rs909735641 Chr.11: 66282116-66282116 on Build GRCh38
rs890819035 Chr.11: 66282154-66282154 on Build GRCh38
rs1011673394 Chr.11: 66282238-66282238 on Build GRCh38
rs773251076 Chr.11: 66282210-66282210 on Build GRCh38

View Full Product Details