Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16552292-16552525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF287
Entrez Gene ID
57336
Gene Name
zinc finger protein 287
Gene Aliases
ZKSCAN13, ZSCAN45
UniGene
Hs.99724
Gene Chromosome Location
Chr.17: 16546689-16569211 on Build GRCh38
Transcripts
NM_020653.2 XM_011523968.2 XM_017024887.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5679521 c.1790G>A p.C597Y Chr.17: 16552352-16552352 on Build GRCh38
975948 c.1676C>T p.S559F Chr.17: 16552466-16552466 on Build GRCh38
4627498 c.1649A>T p.Y550F Chr.17: 16552493-16552493 on Build GRCh38
5000393 c.1730C>T p.S577L Chr.17: 16552412-16552412 on Build GRCh38
975950 c.1667G>A p.R556K Chr.17: 16552475-16552475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766760904 Chr.17: 16552476-16552476 on Build GRCh38
rs1019983850 Chr.17: 16552365-16552365 on Build GRCh38
rs759758331 Chr.17: 16552485-16552485 on Build GRCh38
rs754966099 Chr.17: 16552492-16552492 on Build GRCh38
rs779478641 Chr.17: 16552479-16552479 on Build GRCh38
rs767792852 Chr.17: 16552495-16552495 on Build GRCh38
rs201270094 Chr.17: 16552411-16552411 on Build GRCh38
rs771653161 Chr.17: 16552383-16552383 on Build GRCh38
rs778664389 Chr.17: 16552368-16552368 on Build GRCh38
rs144182359 Chr.17: 16552425-16552425 on Build GRCh38
rs774498338 Chr.17: 16552482-16552482 on Build GRCh38
rs144535422 Chr.17: 16552355-16552355 on Build GRCh38
rs773819985 Chr.17: 16552450-16552450 on Build GRCh38
rs78436090 Chr.17: 16552373-16552373 on Build GRCh38
rs757111179 Chr.17: 16552366-16552366 on Build GRCh38
rs749011167 Chr.17: 16552363-16552363 on Build GRCh38
rs113684848 Chr.17: 16552458-16552458 on Build GRCh38
rs775147680 Chr.17: 16552398-16552398 on Build GRCh38
rs530236422 Chr.17: 16552474-16552474 on Build GRCh38
rs770190449 Chr.17: 16552449-16552449 on Build GRCh38
rs868386276 Chr.17: 16552484-16552484 on Build GRCh38
rs777822571 Chr.17: 16552356-16552356 on Build GRCh38
rs746598310 Chr.17: 16552448-16552448 on Build GRCh38
rs752562155 Chr.17: 16552331-16552331 on Build GRCh38
rs897036095 Chr.17: 16552367-16552367 on Build GRCh38
rs745541947 Chr.17: 16552381-16552381 on Build GRCh38

View Full Product Details