Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26575855-26576126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf70
Entrez Gene ID
339778
Gene Name
chromosome 2 open reading frame 70
Gene Aliases
-
UniGene
Hs.407083
Gene Chromosome Location
Chr.2: 26562559-26581166 on Build GRCh38
Transcripts
NM_001105519.2 NM_001322426.1 XM_011532810.2 XM_011532811.2 XM_017003962.1 XM_017003963.1 XM_017003964.1 XM_017003965.1 XM_017003966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
139380 c.148C>T p.Q50* Chr.2: 26575975-26575975 on Build GRCh38
6615905 c.231delC p.N79fs*4 Chr.2: 26576058-26576058 on Build GRCh38
6615904 c.254G>A p.R85H Chr.2: 26576081-26576081 on Build GRCh38
6615903 c.114C>T p.G38G Chr.2: 26575941-26575941 on Build GRCh38
4001631 c.198A>T p.Q66H Chr.2: 26576025-26576025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753762077 Chr.2: 26575914-26575914 on Build GRCh38
rs759830210 Chr.2: 26575891-26575891 on Build GRCh38
rs771408978 Chr.2: 26575955-26575955 on Build GRCh38
rs376892936 Chr.2: 26575890-26575890 on Build GRCh38
rs367622220 Chr.2: 26575985-26575985 on Build GRCh38
rs1041225425 Chr.2: 26576082-26576082 on Build GRCh38
rs138550936 Chr.2: 26576099-26576099 on Build GRCh38
rs531808422 Chr.2: 26575998-26575998 on Build GRCh38
rs763609891 Chr.2: 26575878-26575878 on Build GRCh38
rs749380596 Chr.2: 26575951-26575951 on Build GRCh38
rs931060743 Chr.2: 26575884-26575884 on Build GRCh38
rs61742175 Chr.2: 26576029-26576029 on Build GRCh38
rs771809134 Chr.2: 26576016-26576016 on Build GRCh38
rs370346487 Chr.2: 26575902-26575902 on Build GRCh38
rs534721835 Chr.2: 26575959-26575959 on Build GRCh38
rs777315280 Chr.2: 26575908-26575908 on Build GRCh38
rs753279641 Chr.2: 26575994-26575994 on Build GRCh38
rs758184713 Chr.2: 26576015-26576015 on Build GRCh38
rs770944861 Chr.2: 26576057-26576057 on Build GRCh38
rs778722509 Chr.2: 26575918-26575918 on Build GRCh38
rs1021612309 Chr.2: 26575928-26575928 on Build GRCh38
rs763590692 Chr.2: 26575984-26575984 on Build GRCh38
rs181202795 Chr.2: 26575919-26575919 on Build GRCh38
rs778151436 Chr.2: 26576028-26576028 on Build GRCh38
rs201740466 Chr.2: 26576093-26576093 on Build GRCh38
rs866402113 Chr.2: 26576052-26576052 on Build GRCh38
rs767148167 Chr.2: 26575896-26575896 on Build GRCh38
rs758331071 Chr.2: 26576098-26576098 on Build GRCh38
rs567506070 Chr.2: 26575941-26575941 on Build GRCh38
rs376790545 Chr.2: 26576080-26576080 on Build GRCh38
rs969144882 Chr.2: 26575938-26575938 on Build GRCh38
rs756944732 Chr.2: 26575929-26575931 on Build GRCh38
rs764698232 Chr.2: 26576092-26576092 on Build GRCh38
rs759903840 Chr.2: 26575961-26575961 on Build GRCh38
rs780049642 Chr.2: 26576019-26576019 on Build GRCh38
rs766644577 Chr.2: 26575885-26575885 on Build GRCh38
rs572327407 Chr.2: 26576083-26576083 on Build GRCh38
rs763680722 Chr.2: 26576081-26576081 on Build GRCh38
rs764726900 Chr.2: 26576075-26576076 on Build GRCh38
rs542880558 Chr.2: 26576088-26576088 on Build GRCh38
rs112873448 Chr.2: 26576059-26576059 on Build GRCh38
rs767810976 Chr.2: 26575978-26575978 on Build GRCh38
rs748424847 Chr.2: 26575945-26575945 on Build GRCh38
rs776830826 Chr.2: 26575897-26575897 on Build GRCh38
rs553972095 Chr.2: 26576076-26576076 on Build GRCh38
rs754839442 Chr.2: 26576108-26576108 on Build GRCh38
rs561356267 Chr.2: 26575942-26575942 on Build GRCh38
rs13002673 Chr.2: 26576025-26576025 on Build GRCh38
rs760379580 Chr.2: 26575979-26575979 on Build GRCh38
rs201943227 Chr.2: 26575903-26575903 on Build GRCh38
rs374971635 Chr.2: 26575915-26575915 on Build GRCh38
rs751338382 Chr.2: 26576106-26576106 on Build GRCh38
rs749494116 Chr.2: 26575950-26575950 on Build GRCh38
rs765175546 Chr.2: 26576004-26576004 on Build GRCh38
rs372202665 Chr.2: 26576006-26576006 on Build GRCh38
rs772471808 Chr.2: 26576073-26576073 on Build GRCh38

View Full Product Details