Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64414816-64415077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XPOT
Entrez Gene ID
11260
Gene Name
exportin for tRNA
Gene Aliases
XPO3
UniGene
Hs.592579
Gene Chromosome Location
Chr.12: 64404350-64451125 on Build GRCh38
Transcripts
NM_007235.4 XM_017018748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6824100 c.89_90insC p.S31fs*19 Chr.12: 64414935-64414936 on Build GRCh38
7201626 c.61-22_61-9delTTTTTGTTTTGCAA p.? Chr.12: 64414885-64414898 on Build GRCh38
4044149 c.62C>T p.A21V Chr.12: 64414908-64414908 on Build GRCh38
3464215 c.92C>T p.S31F Chr.12: 64414938-64414938 on Build GRCh38
431660 c.132G>A p.Q44Q Chr.12: 64414978-64414978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763821493 Chr.12: 64414915-64414915 on Build GRCh38
rs777498205 Chr.12: 64414864-64414864 on Build GRCh38
rs777618918 Chr.12: 64414995-64414996 on Build GRCh38
rs772131319 Chr.12: 64414904-64414904 on Build GRCh38
rs758473702 Chr.12: 64414958-64414958 on Build GRCh38
rs1053379137 Chr.12: 64415009-64415009 on Build GRCh38
rs777434002 Chr.12: 64414884-64414884 on Build GRCh38
rs761994641 Chr.12: 64414934-64414934 on Build GRCh38
rs370935460 Chr.12: 64414951-64414951 on Build GRCh38
rs370184054 Chr.12: 64414899-64414899 on Build GRCh38
rs144596187 Chr.12: 64414963-64414963 on Build GRCh38
rs559679683 Chr.12: 64414876-64414876 on Build GRCh38
rs765498017 Chr.12: 64414938-64414938 on Build GRCh38
rs748593229 Chr.12: 64414996-64414996 on Build GRCh38
rs752027736 Chr.12: 64414970-64414970 on Build GRCh38
rs781378289 Chr.12: 64414988-64414988 on Build GRCh38
rs199897284 Chr.12: 64415032-64415032 on Build GRCh38
rs200678967 Chr.12: 64415001-64415001 on Build GRCh38
rs769132232 Chr.12: 64414882-64414883 on Build GRCh38
rs776875305 Chr.12: 64414928-64414928 on Build GRCh38
rs768912800 Chr.12: 64414882-64414882 on Build GRCh38
rs150255616 Chr.12: 64414909-64414909 on Build GRCh38
rs368006228 Chr.12: 64414895-64414895 on Build GRCh38
rs375213242 Chr.12: 64415015-64415015 on Build GRCh38
rs747328249 Chr.12: 64414881-64414881 on Build GRCh38
rs755329576 Chr.12: 64414979-64414979 on Build GRCh38
rs760464151 Chr.12: 64414914-64414914 on Build GRCh38
rs777561905 Chr.12: 64414969-64414969 on Build GRCh38
rs771722448 Chr.12: 64415038-64415038 on Build GRCh38
rs532716871 Chr.12: 64415012-64415012 on Build GRCh38
rs368225863 Chr.12: 64415031-64415031 on Build GRCh38
rs1039252367 Chr.12: 64414845-64414845 on Build GRCh38

View Full Product Details