Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99763876-99764101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP3A4
Entrez Gene ID
1576
Gene Name
cytochrome P450 family 3 subfamily A member 4
Gene Aliases
CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, HLP, NF-25, P450C3, P450PCN1
UniGene
Hs.728751
Gene Chromosome Location
Chr.7: 99756960-99784188 on Build GRCh38
Transcripts
NM_001202855.2 NM_017460.5 XM_017011780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4993042 c.944C>T p.S315F Chr.7: 99763937-99763937 on Build GRCh38
3883883 c.889G>A p.A297T Chr.7: 99763992-99763992 on Build GRCh38
1093882 c.942C>A p.L314L Chr.7: 99763939-99763939 on Build GRCh38
6331037 c.954G>A p.M318I Chr.7: 99763927-99763927 on Build GRCh38
1699001 c.958G>A p.E320K Chr.7: 99763923-99763923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201450467 Chr.7: 99763938-99763938 on Build GRCh38
rs770129614 Chr.7: 99763937-99763937 on Build GRCh38
rs756077272 Chr.7: 99763986-99763986 on Build GRCh38
rs766060602 Chr.7: 99763949-99763949 on Build GRCh38
rs145779318 Chr.7: 99763951-99763951 on Build GRCh38
rs765103065 Chr.7: 99764009-99764009 on Build GRCh38
rs773937044 Chr.7: 99764050-99764050 on Build GRCh38
rs540702483 Chr.7: 99764007-99764007 on Build GRCh38
rs753982121 Chr.7: 99764029-99764029 on Build GRCh38
rs71581996 Chr.7: 99763968-99763968 on Build GRCh38
rs375997724 Chr.7: 99763955-99763955 on Build GRCh38
rs201821708 Chr.7: 99763925-99763925 on Build GRCh38
rs753534506 Chr.7: 99763915-99763917 on Build GRCh38
rs1015541328 Chr.7: 99763939-99763939 on Build GRCh38
rs190354371 Chr.7: 99763940-99763940 on Build GRCh38
rs143158202 Chr.7: 99763990-99763990 on Build GRCh38
rs773595509 Chr.7: 99764035-99764035 on Build GRCh38
rs758999970 Chr.7: 99763956-99763956 on Build GRCh38
rs771309969 Chr.7: 99764045-99764045 on Build GRCh38
rs368676034 Chr.7: 99764041-99764041 on Build GRCh38
rs774686251 Chr.7: 99764005-99764005 on Build GRCh38
rs71581997 Chr.7: 99763960-99763960 on Build GRCh38
rs751246524 Chr.7: 99763952-99763952 on Build GRCh38
rs1056231533 Chr.7: 99763898-99763898 on Build GRCh38
rs187260898 Chr.7: 99764008-99764008 on Build GRCh38
rs138675831 Chr.7: 99763995-99763995 on Build GRCh38
rs777614318 Chr.7: 99763987-99763987 on Build GRCh38
rs762572678 Chr.7: 99763943-99763943 on Build GRCh38
rs150187589 Chr.7: 99763920-99763920 on Build GRCh38
rs368465788 Chr.7: 99763996-99763996 on Build GRCh38
rs267601666 Chr.7: 99763941-99763941 on Build GRCh38
rs201286895 Chr.7: 99763983-99763983 on Build GRCh38
rs28371759 Chr.7: 99764003-99764003 on Build GRCh38

View Full Product Details