Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102229761-102229986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YAP1
Entrez Gene ID
10413
Gene Name
Yes associated protein 1
Gene Aliases
COB1, YAP, YAP2, YAP65, YKI
UniGene
Hs.503692
Gene Chromosome Location
Chr.11: 102109957-102233423 on Build GRCh38
Transcripts
NM_001130145.2 NM_001195044.1 NM_001195045.1 NM_001282097.1 NM_001282098.1 NM_001282099.1 NM_001282100.1 NM_001282101.1 NM_006106.4 XM_005271378.3 XM_005271380.3 XM_005271381.3 XM_005271383.3 XM_011542555.2 XM_011542556.2 XM_017017093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6952281 c.1501C>A p.L501I Chr.11: 102229926-102229926 on Build GRCh38
5802533 c.1486delG p.D496fs*>9 Chr.11: 102229911-102229911 on Build GRCh38
4165611 c.1477A>G p.T493A Chr.11: 102229902-102229902 on Build GRCh38
3985859 c.1237G>T p.G413* Chr.11: 102229824-102229824 on Build GRCh38
3791100 c.1248G>C p.L416L Chr.11: 102229835-102229835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201772622 Chr.11: 102229818-102229818 on Build GRCh38
rs369837465 Chr.11: 102229952-102229952 on Build GRCh38
rs773457076 Chr.11: 102229785-102229785 on Build GRCh38
rs1022838674 Chr.11: 102229828-102229828 on Build GRCh38
rs112713899 Chr.11: 102229871-102229871 on Build GRCh38
rs779614719 Chr.11: 102229937-102229937 on Build GRCh38
rs759882394 Chr.11: 102229829-102229829 on Build GRCh38
rs754789933 Chr.11: 102229951-102229951 on Build GRCh38
rs756621827 Chr.11: 102229861-102229861 on Build GRCh38
rs758047587 Chr.11: 102229919-102229919 on Build GRCh38
rs769873665 Chr.11: 102229953-102229953 on Build GRCh38
rs749962892 Chr.11: 102229902-102229902 on Build GRCh38
rs534468390 Chr.11: 102229844-102229844 on Build GRCh38
rs148574447 Chr.11: 102229821-102229821 on Build GRCh38
rs746816956 Chr.11: 102229945-102229945 on Build GRCh38
rs766554722 Chr.11: 102229819-102229819 on Build GRCh38
rs759000569 Chr.11: 102229934-102229935 on Build GRCh38
rs141269128 Chr.11: 102229841-102229841 on Build GRCh38
rs764690657 Chr.11: 102229884-102229884 on Build GRCh38
rs956054285 Chr.11: 102229947-102229947 on Build GRCh38

View Full Product Details