Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150267246-150267507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APH1A
Entrez Gene ID
51107
Gene Name
aph-1 homolog A, gamma-secretase subunit
Gene Aliases
6530402N02Rik, APH-1, APH-1A, CGI-78
UniGene
Hs.108408
Gene Chromosome Location
Chr.1: 150265399-150282291 on Build GRCh38
Transcripts
NM_001077628.2 NM_001243771.1 NM_001243772.1 NM_016022.3 NR_045033.1 NR_045034.1 NR_045035.1 XM_017001417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6602139 c.376G>A p.G126S Chr.1: 150267461-150267461 on Build GRCh38
4496281 c.471C>T p.F157F Chr.1: 150267366-150267366 on Build GRCh38
6602143 c.388G>T p.G130C Chr.1: 150267449-150267449 on Build GRCh38
4496280 c.471C>T p.F157F Chr.1: 150267366-150267366 on Build GRCh38
3976101 c.379A>G p.I127V Chr.1: 150267458-150267458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782508662 Chr.1: 150267365-150267365 on Build GRCh38
rs201704984 Chr.1: 150267344-150267344 on Build GRCh38
rs1013617055 Chr.1: 150267393-150267393 on Build GRCh38
rs201674071 Chr.1: 150267312-150267312 on Build GRCh38
rs782399735 Chr.1: 150267308-150267308 on Build GRCh38
rs140561586 Chr.1: 150267322-150267322 on Build GRCh38
rs781915604 Chr.1: 150267286-150267286 on Build GRCh38
rs764825069 Chr.1: 150267265-150267265 on Build GRCh38
rs782645515 Chr.1: 150267368-150267368 on Build GRCh38
rs200097244 Chr.1: 150267324-150267324 on Build GRCh38
rs375410261 Chr.1: 150267408-150267408 on Build GRCh38
rs200723256 Chr.1: 150267340-150267340 on Build GRCh38
rs753052261 Chr.1: 150267348-150267348 on Build GRCh38
rs199613283 Chr.1: 150267317-150267317 on Build GRCh38
rs148296801 Chr.1: 150267289-150267289 on Build GRCh38
rs201257450 Chr.1: 150267462-150267462 on Build GRCh38
rs1024793009 Chr.1: 150267474-150267474 on Build GRCh38
rs891284194 Chr.1: 150267278-150267278 on Build GRCh38
rs782191839 Chr.1: 150267483-150267483 on Build GRCh38
rs201631567 Chr.1: 150267323-150267323 on Build GRCh38
rs1015865086 Chr.1: 150267295-150267295 on Build GRCh38
rs782572007 Chr.1: 150267433-150267433 on Build GRCh38
rs782486878 Chr.1: 150267381-150267381 on Build GRCh38
rs782124083 Chr.1: 150267325-150267325 on Build GRCh38
rs113330322 Chr.1: 150267355-150267355 on Build GRCh38
rs782216665 Chr.1: 150267378-150267378 on Build GRCh38
rs199893609 Chr.1: 150267345-150267345 on Build GRCh38

View Full Product Details