Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76323163-76323408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC158
Entrez Gene ID
339965
Gene Name
coiled-coil domain containing 158
Gene Aliases
-
UniGene
Hs.529680
Gene Chromosome Location
Chr.4: 76313039-76421920 on Build GRCh38
Transcripts
NM_001042784.1 XM_005262974.4 XM_006714204.3 XM_011531908.2 XM_011531909.2 XM_011531910.2 XM_011531911.1 XM_011531912.2 XM_011531913.1 XM_011531914.2 XM_011531915.2 XM_011531916.2 XM_011531917.1 XM_017008160.1 XM_017008161.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3975029 c.3176T>C p.I1059T Chr.4: 76323391-76323391 on Build GRCh38
5887853 c.3223A>G p.S1075G Chr.4: 76323344-76323344 on Build GRCh38
6621796 c.3171G>A p.P1057P Chr.4: 76323396-76323396 on Build GRCh38
1057658 c.3224G>A p.S1075N Chr.4: 76323343-76323343 on Build GRCh38
6990486 c.3258G>C p.K1086N Chr.4: 76323309-76323309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201014339 Chr.4: 76323336-76323336 on Build GRCh38
rs745357903 Chr.4: 76323272-76323272 on Build GRCh38
rs367816199 Chr.4: 76323270-76323270 on Build GRCh38
rs749047744 Chr.4: 76323265-76323265 on Build GRCh38
rs777600776 Chr.4: 76323242-76323242 on Build GRCh38
rs763332500 Chr.4: 76323327-76323327 on Build GRCh38
rs751888813 Chr.4: 76323352-76323352 on Build GRCh38
rs753073932 Chr.4: 76323251-76323251 on Build GRCh38
rs375934154 Chr.4: 76323313-76323313 on Build GRCh38
rs771256504 Chr.4: 76323310-76323310 on Build GRCh38
rs372030998 Chr.4: 76323357-76323357 on Build GRCh38
rs369670339 Chr.4: 76323326-76323326 on Build GRCh38
rs756948485 Chr.4: 76323353-76323353 on Build GRCh38
rs372733100 Chr.4: 76323304-76323304 on Build GRCh38
rs765671136 Chr.4: 76323323-76323323 on Build GRCh38
rs374833359 Chr.4: 76323269-76323269 on Build GRCh38
rs180974490 Chr.4: 76323240-76323240 on Build GRCh38
rs777418292 Chr.4: 76323266-76323266 on Build GRCh38
rs780036058 Chr.4: 76323277-76323277 on Build GRCh38
rs776634466 Chr.4: 76323312-76323312 on Build GRCh38
rs773600556 Chr.4: 76323301-76323301 on Build GRCh38
rs757231081 Chr.4: 76323198-76323198 on Build GRCh38
rs930062105 Chr.4: 76323371-76323371 on Build GRCh38
rs749039685 Chr.4: 76323291-76323291 on Build GRCh38
rs769677735 Chr.4: 76323273-76323273 on Build GRCh38

View Full Product Details