Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11831496-11831751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF440
Entrez Gene ID
126070
Gene Name
zinc finger protein 440
Gene Aliases
-
UniGene
Hs.418192
Gene Chromosome Location
Chr.19: 11814241-11835617 on Build GRCh38
Transcripts
NM_152357.2 XM_005259731.3 XM_017026253.1 XM_017026254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4570916 c.347T>G p.L116R Chr.19: 11831523-11831523 on Build GRCh38
5818884 c.367A>G p.M123V Chr.19: 11831543-11831543 on Build GRCh38
4517895 c.455_456CC>TT p.A152V Chr.19: 11831631-11831632 on Build GRCh38
4550652 c.505G>A p.E169K Chr.19: 11831681-11831681 on Build GRCh38
1390457 c.531T>C p.C177C Chr.19: 11831707-11831707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369098092 Chr.19: 11831677-11831677 on Build GRCh38
rs751383865 Chr.19: 11831572-11831572 on Build GRCh38
rs753068694 Chr.19: 11831663-11831663 on Build GRCh38
rs757006207 Chr.19: 11831584-11831607 on Build GRCh38
rs796157809 Chr.19: 11831655-11831655 on Build GRCh38
rs745789303 Chr.19: 11831694-11831694 on Build GRCh38
rs920133183 Chr.19: 11831608-11831608 on Build GRCh38
rs1057213207 Chr.19: 11831647-11831647 on Build GRCh38
rs1000821042 Chr.19: 11831708-11831708 on Build GRCh38
rs564798619 Chr.19: 11831594-11831594 on Build GRCh38
rs752036599 Chr.19: 11831653-11831653 on Build GRCh38
rs559327841 Chr.19: 11831598-11831598 on Build GRCh38
rs427880 Chr.19: 11831547-11831547 on Build GRCh38
rs937237987 Chr.19: 11831632-11831632 on Build GRCh38
rs912056812 Chr.19: 11831532-11831532 on Build GRCh38
rs777650792 Chr.19: 11831585-11831585 on Build GRCh38
rs375155828 Chr.19: 11831712-11831712 on Build GRCh38
rs200883112 Chr.19: 11831621-11831621 on Build GRCh38
rs752946742 Chr.19: 11831644-11831644 on Build GRCh38
rs753237656 Chr.19: 11831638-11831638 on Build GRCh38
rs781582316 Chr.19: 11831559-11831559 on Build GRCh38
rs770725775 Chr.19: 11831599-11831599 on Build GRCh38
rs768154473 Chr.19: 11831704-11831704 on Build GRCh38
rs894575178 Chr.19: 11831652-11831652 on Build GRCh38
rs200222542 Chr.19: 11831643-11831643 on Build GRCh38
rs755301419 Chr.19: 11831548-11831548 on Build GRCh38
rs368640946 Chr.19: 11831541-11831541 on Build GRCh38
rs770071474 Chr.19: 11831575-11831575 on Build GRCh38
rs367654656 Chr.19: 11831561-11831561 on Build GRCh38
rs771971997 Chr.19: 11831620-11831620 on Build GRCh38
rs780633623 Chr.19: 11831546-11831546 on Build GRCh38
rs761412705 Chr.19: 11831641-11831641 on Build GRCh38
rs777294336 Chr.19: 11831710-11831710 on Build GRCh38
rs760335822 Chr.19: 11831634-11831634 on Build GRCh38
rs201981931 Chr.19: 11831642-11831642 on Build GRCh38
rs771012865 Chr.19: 11831690-11831690 on Build GRCh38
rs755534063 Chr.19: 11831660-11831660 on Build GRCh38
rs931621659 Chr.19: 11831612-11831612 on Build GRCh38
rs757987981 Chr.19: 11831688-11831688 on Build GRCh38
rs774378462 Chr.19: 11831616-11831616 on Build GRCh38
rs760568774 Chr.19: 11831703-11831703 on Build GRCh38
rs200311015 Chr.19: 11831618-11831618 on Build GRCh38
rs778691104 Chr.19: 11831562-11831562 on Build GRCh38
rs749288817 Chr.19: 11831681-11831681 on Build GRCh38
rs756237026 Chr.19: 11831665-11831665 on Build GRCh38
rs781535111 Chr.19: 11831661-11831661 on Build GRCh38
rs944914486 Chr.19: 11831543-11831543 on Build GRCh38
rs576726445 Chr.19: 11831544-11831544 on Build GRCh38
rs371957789 Chr.19: 11831698-11831698 on Build GRCh38
rs763505241 Chr.19: 11831525-11831525 on Build GRCh38
rs763820178 Chr.19: 11831635-11831635 on Build GRCh38
rs772139200 Chr.19: 11831697-11831697 on Build GRCh38
rs753391461 Chr.19: 11831526-11831526 on Build GRCh38
rs754886559 Chr.19: 11831557-11831557 on Build GRCh38
rs778910663 Chr.19: 11831691-11831691 on Build GRCh38
rs763684851 Chr.19: 11831550-11831551 on Build GRCh38

View Full Product Details