Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78997168-78997405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CANT1
Entrez Gene ID
124583
Gene Name
calcium activated nucleotidase 1
Gene Aliases
DBQD, DBQD1, SCAN-1, SCAN1, SHAPY
UniGene
Hs.8859
Gene Chromosome Location
Chr.17: 78991716-79009817 on Build GRCh38
Transcripts
NM_001159772.1 NM_001159773.1 NM_138793.3 XM_005257021.1 XM_005257022.1 XM_006721683.1 XM_011524291.1 XM_011524293.1 XM_011524294.1 XM_011524295.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6996374 c.398A>G p.Y133C Chr.17: 78997225-78997225 on Build GRCh38
2746870 c.392delA p.K131fs*4 Chr.17: 78997231-78997231 on Build GRCh38
2746872 c.305G>T p.G102V Chr.17: 78997318-78997318 on Build GRCh38
6894723 c.402G>A p.L134L Chr.17: 78997221-78997221 on Build GRCh38
5853667 c.346G>A p.E116K Chr.17: 78997277-78997277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567995164 Chr.17: 78997377-78997377 on Build GRCh38
rs771723898 Chr.17: 78997191-78997191 on Build GRCh38
rs753599330 Chr.17: 78997339-78997339 on Build GRCh38
rs776925548 Chr.17: 78997196-78997196 on Build GRCh38
rs759677564 Chr.17: 78997329-78997329 on Build GRCh38
rs763510198 Chr.17: 78997210-78997210 on Build GRCh38
rs148785781 Chr.17: 78997306-78997306 on Build GRCh38
rs587776898 Chr.17: 78997248-78997248 on Build GRCh38
rs768167035 Chr.17: 78997355-78997355 on Build GRCh38
rs773784258 Chr.17: 78997357-78997357 on Build GRCh38
rs966594441 Chr.17: 78997243-78997243 on Build GRCh38
rs760997716 Chr.17: 78997375-78997375 on Build GRCh38
rs749661486 Chr.17: 78997361-78997361 on Build GRCh38
rs980361277 Chr.17: 78997291-78997291 on Build GRCh38
rs767601069 Chr.17: 78997345-78997345 on Build GRCh38
rs775411325 Chr.17: 78997374-78997374 on Build GRCh38
rs757055426 Chr.17: 78997385-78997385 on Build GRCh38
rs774522276 Chr.17: 78997275-78997276 on Build GRCh38
rs751663458 Chr.17: 78997223-78997223 on Build GRCh38
rs778807955 Chr.17: 78997345-78997345 on Build GRCh38
rs988911903 Chr.17: 78997235-78997235 on Build GRCh38
rs752806482 Chr.17: 78997265-78997265 on Build GRCh38
rs369008128 Chr.17: 78997350-78997350 on Build GRCh38
rs752996676 Chr.17: 78997332-78997332 on Build GRCh38
rs757164885 Chr.17: 78997292-78997292 on Build GRCh38
rs200503476 Chr.17: 78997313-78997313 on Build GRCh38
rs755322718 Chr.17: 78997239-78997239 on Build GRCh38
rs267606702 Chr.17: 78997249-78997249 on Build GRCh38
rs140112462 Chr.17: 78997190-78997190 on Build GRCh38
rs372736806 Chr.17: 78997354-78997354 on Build GRCh38
rs759923329 Chr.17: 78997371-78997371 on Build GRCh38
rs777393447 Chr.17: 78997286-78997286 on Build GRCh38
rs867789182 Chr.17: 78997198-78997198 on Build GRCh38
rs775372185 Chr.17: 78997323-78997323 on Build GRCh38
rs61738470 Chr.17: 78997250-78997250 on Build GRCh38
rs781465433 Chr.17: 78997258-78997258 on Build GRCh38
rs147336056 Chr.17: 78997201-78997201 on Build GRCh38
rs763599455 Chr.17: 78997312-78997312 on Build GRCh38
rs771367044 Chr.17: 78997364-78997364 on Build GRCh38
rs372332465 Chr.17: 78997307-78997307 on Build GRCh38
rs199762751 Chr.17: 78997365-78997365 on Build GRCh38
rs773657859 Chr.17: 78997311-78997311 on Build GRCh38
rs779389320 Chr.17: 78997352-78997352 on Build GRCh38
rs932427636 Chr.17: 78997309-78997309 on Build GRCh38
rs755943793 Chr.17: 78997271-78997271 on Build GRCh38
rs909608446 Chr.17: 78997338-78997338 on Build GRCh38
rs771678393 Chr.17: 78997295-78997295 on Build GRCh38
rs749246739 Chr.17: 78997287-78997287 on Build GRCh38
rs757251090 Chr.17: 78997344-78997344 on Build GRCh38
rs764942510 Chr.17: 78997386-78997386 on Build GRCh38
rs766584428 Chr.17: 78997216-78997216 on Build GRCh38
rs587776897 Chr.17: 78997345-78997346 on Build GRCh38
rs368963278 Chr.17: 78997293-78997293 on Build GRCh38
rs775218418 Chr.17: 78997302-78997302 on Build GRCh38
rs976307747 Chr.17: 78997244-78997244 on Build GRCh38
rs535302434 Chr.17: 78997204-78997204 on Build GRCh38
rs375718883 Chr.17: 78997336-78997336 on Build GRCh38
rs142062082 Chr.17: 78997378-78997378 on Build GRCh38
rs745377146 Chr.17: 78997294-78997294 on Build GRCh38
rs767659113 Chr.17: 78997373-78997373 on Build GRCh38
rs767430805 Chr.17: 78997330-78997330 on Build GRCh38

View Full Product Details