Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 742709-742970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YES1
Entrez Gene ID
7525
Gene Name
YES proto-oncogene 1, Src family tyrosine kinase
Gene Aliases
HsT441, P61-YES, Yes, c-yes
UniGene
Hs.194148
Gene Chromosome Location
Chr.18: 721592-812326 on Build GRCh38
Transcripts
NM_005433.3 XM_005258139.3 XM_017025960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4818697 c.1034A>C p.Y345S Chr.18: 742944-742944 on Build GRCh38
294075 c.1027C>T p.P343S Chr.18: 742951-742951 on Build GRCh38
6915720 c.1057A>G p.K353E Chr.18: 742921-742921 on Build GRCh38
6959095 c.1043C>T p.T348I Chr.18: 742935-742935 on Build GRCh38
6912334 c.1019C>G p.S340C Chr.18: 742959-742959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780838371 Chr.18: 742935-742935 on Build GRCh38
rs753062108 Chr.18: 742918-742918 on Build GRCh38
rs766635490 Chr.18: 742887-742887 on Build GRCh38
rs368921388 Chr.18: 742869-742869 on Build GRCh38
rs765484832 Chr.18: 742916-742916 on Build GRCh38
rs887564337 Chr.18: 742733-742733 on Build GRCh38
rs760775133 Chr.18: 742886-742886 on Build GRCh38
rs773323129 Chr.18: 742884-742884 on Build GRCh38
rs114740824 Chr.18: 742891-742891 on Build GRCh38
rs755587747 Chr.18: 742903-742903 on Build GRCh38
rs9961743 Chr.18: 742858-742858 on Build GRCh38
rs960481111 Chr.18: 742779-742779 on Build GRCh38
rs1033887118 Chr.18: 742743-742743 on Build GRCh38
rs758709210 Chr.18: 742921-742921 on Build GRCh38
rs370426200 Chr.18: 742841-742841 on Build GRCh38
rs761544020 Chr.18: 742870-742870 on Build GRCh38
rs547798212 Chr.18: 742864-742864 on Build GRCh38
rs1014462100 Chr.18: 742838-742838 on Build GRCh38
rs767730645 Chr.18: 742875-742875 on Build GRCh38

View Full Product Details