Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 20626450-20626707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB5
Entrez Gene ID
340273
Gene Name
ATP binding cassette subfamily B member 5
Gene Aliases
ABCB5alpha, ABCB5beta, EST422562
UniGene
Hs.404102
Gene Chromosome Location
Chr.7: 20615622-20757014 on Build GRCh38
Transcripts
NM_001163941.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5815964 c.58G>A p.A20T Chr.7: 20626561-20626561 on Build GRCh38
5459862 c.108+1G>C p.? Chr.7: 20626612-20626612 on Build GRCh38
5407211 c.85G>A p.E29K Chr.7: 20626588-20626588 on Build GRCh38
6109810 c.69G>T p.Q23H Chr.7: 20626572-20626572 on Build GRCh38
1449893 c.92T>A p.V31D Chr.7: 20626595-20626595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563029385 Chr.7: 20626567-20626567 on Build GRCh38
rs746232251 Chr.7: 20626628-20626629 on Build GRCh38
rs1004187349 Chr.7: 20626633-20626633 on Build GRCh38
rs558983048 Chr.7: 20626598-20626598 on Build GRCh38
rs368439981 Chr.7: 20626604-20626604 on Build GRCh38
rs566413954 Chr.7: 20626500-20626500 on Build GRCh38
rs375221629 Chr.7: 20626528-20626528 on Build GRCh38
rs752049509 Chr.7: 20626558-20626558 on Build GRCh38
rs764215526 Chr.7: 20626613-20626613 on Build GRCh38
rs536940255 Chr.7: 20626503-20626503 on Build GRCh38
rs370748202 Chr.7: 20626614-20626614 on Build GRCh38
rs1037484282 Chr.7: 20626662-20626662 on Build GRCh38
rs771442695 Chr.7: 20626532-20626532 on Build GRCh38
rs761794408 Chr.7: 20626595-20626595 on Build GRCh38
rs776107227 Chr.7: 20626570-20626570 on Build GRCh38
rs761103435 Chr.7: 20626518-20626518 on Build GRCh38
rs957672966 Chr.7: 20626534-20626534 on Build GRCh38
rs757909451 Chr.7: 20626624-20626624 on Build GRCh38
rs576480452 Chr.7: 20626502-20626502 on Build GRCh38
rs1012222883 Chr.7: 20626555-20626555 on Build GRCh38
rs765395888 Chr.7: 20626609-20626609 on Build GRCh38
rs758498909 Chr.7: 20626536-20626536 on Build GRCh38
rs768087628 Chr.7: 20626585-20626585 on Build GRCh38
rs752344297 Chr.7: 20626642-20626642 on Build GRCh38
rs182947689 Chr.7: 20626631-20626631 on Build GRCh38
rs991442570 Chr.7: 20626487-20626487 on Build GRCh38
rs376372685 Chr.7: 20626579-20626579 on Build GRCh38
rs750803775 Chr.7: 20626635-20626635 on Build GRCh38
rs140851914 Chr.7: 20626568-20626568 on Build GRCh38
rs373223407 Chr.7: 20626574-20626574 on Build GRCh38
rs756453071 Chr.7: 20626643-20626643 on Build GRCh38
rs373464355 Chr.7: 20626625-20626625 on Build GRCh38
rs762691730 Chr.7: 20626647-20626647 on Build GRCh38
rs778451850 Chr.7: 20626551-20626551 on Build GRCh38
rs765814085 Chr.7: 20626563-20626565 on Build GRCh38
rs201244034 Chr.7: 20626603-20626603 on Build GRCh38
rs187496867 Chr.7: 20626674-20626674 on Build GRCh38
rs78309031 Chr.7: 20626562-20626562 on Build GRCh38
rs901563978 Chr.7: 20626636-20626636 on Build GRCh38
rs543570107 Chr.7: 20626590-20626590 on Build GRCh38
rs369958537 Chr.7: 20626583-20626583 on Build GRCh38
rs17143187 Chr.7: 20626612-20626612 on Build GRCh38
rs779604830 Chr.7: 20626648-20626648 on Build GRCh38
rs910225145 Chr.7: 20626617-20626617 on Build GRCh38
rs949730989 Chr.7: 20626522-20626522 on Build GRCh38
rs1053313481 Chr.7: 20626672-20626672 on Build GRCh38
rs1049441253 Chr.7: 20626616-20626616 on Build GRCh38

View Full Product Details