Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19884230-19884483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHB
Entrez Gene ID
151651
Gene Name
EF-hand domain family member B
Gene Aliases
CFAP21
UniGene
Hs.733920
Gene Chromosome Location
Chr.3: 19879472-19947036 on Build GRCh38
Transcripts
NM_144715.3 XM_005264889.1 XM_011533382.1 XM_011533383.2 XM_011533385.2 XM_017005741.1 XM_017005743.1 XM_017005744.1 XM_017005745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4789401 c.2070A>G p.P690P Chr.3: 19884479-19884479 on Build GRCh38
4840415 c.2112G>T p.E704D Chr.3: 19884437-19884437 on Build GRCh38
1043241 c.2100A>G p.T700T Chr.3: 19884449-19884449 on Build GRCh38
272270 c.2089T>C p.Y697H Chr.3: 19884460-19884460 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs907194412 Chr.3: 19884448-19884448 on Build GRCh38
rs925824766 Chr.3: 19884332-19884332 on Build GRCh38
rs570902003 Chr.3: 19884321-19884321 on Build GRCh38
rs117124787 Chr.3: 19884275-19884275 on Build GRCh38
rs560529462 Chr.3: 19884440-19884440 on Build GRCh38
rs773453676 Chr.3: 19884432-19884432 on Build GRCh38
rs985789665 Chr.3: 19884354-19884354 on Build GRCh38
rs992675778 Chr.3: 19884302-19884302 on Build GRCh38
rs9882578 Chr.3: 19884315-19884315 on Build GRCh38
rs368077923 Chr.3: 19884360-19884360 on Build GRCh38
rs766850033 Chr.3: 19884441-19884441 on Build GRCh38
rs1006490302 Chr.3: 19884261-19884261 on Build GRCh38
rs774923449 Chr.3: 19884357-19884357 on Build GRCh38
rs936145612 Chr.3: 19884331-19884331 on Build GRCh38
rs994381193 Chr.3: 19884387-19884387 on Build GRCh38
rs922944908 Chr.3: 19884292-19884292 on Build GRCh38
rs975741627 Chr.3: 19884287-19884287 on Build GRCh38
rs140270457 Chr.3: 19884345-19884345 on Build GRCh38
rs755147016 Chr.3: 19884450-19884450 on Build GRCh38
rs147936472 Chr.3: 19884449-19884449 on Build GRCh38
rs765596488 Chr.3: 19884420-19884420 on Build GRCh38
rs776312787 Chr.3: 19884402-19884402 on Build GRCh38
rs771911716 Chr.3: 19884355-19884355 on Build GRCh38
rs771425068 Chr.3: 19884388-19884391 on Build GRCh38
rs966390953 Chr.3: 19884308-19884308 on Build GRCh38
rs948987386 Chr.3: 19884365-19884365 on Build GRCh38
rs1017498551 Chr.3: 19884263-19884263 on Build GRCh38
rs761392775 Chr.3: 19884419-19884419 on Build GRCh38
rs768295969 Chr.3: 19884389-19884389 on Build GRCh38

View Full Product Details