Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21382843-21383098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIPEPP3
Entrez Gene ID
650794
Gene Name
mitochondrial intermediate peptidase pseudogene 3
Gene Aliases
-
UniGene
Hs.147563
Gene Chromosome Location
Chr.13: 21298125-21392923 on Build GRCh38
Transcripts
NR_046461.1 NM_001286638.1 NM_153251.3 NR_104486.1 NR_104487.1 XM_006719795.1 XM_006719796.1 XM_006719797.1 XM_011535030.2 XM_017020525.1 XM_017020526.1 XM_017020527.1

Gene Symbol
ZDHHC20
Entrez Gene ID
253832
Gene Name
zinc finger DHHC-type containing 20
Gene Aliases
4933421L13Rik, DHHC-20
UniGene
Hs.564611
Gene Chromosome Location
Chr.13: 21372571-21459370 on Build GRCh38
Transcripts
NR_046461.1 NM_001286638.1 NM_153251.3 NR_104486.1 NR_104487.1 XM_006719795.1 XM_006719796.1 XM_006719797.1 XM_011535030.2 XM_017020525.1 XM_017020526.1 XM_017020527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6364744 c.906A>T p.E302D Chr.13: 21382958-21382958 on Build GRCh38
3813556 c.904G>A p.E302K Chr.13: 21382960-21382960 on Build GRCh38
3813557 c.904G>A p.E302K Chr.13: 21382960-21382960 on Build GRCh38
7398409 c.883C>T p.R295C Chr.13: 21382981-21382981 on Build GRCh38
3813555 c.904G>A p.E302K Chr.13: 21382960-21382960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953456256 Chr.13: 21382917-21382917 on Build GRCh38
rs894139589 Chr.13: 21383045-21383045 on Build GRCh38
rs779083613 Chr.13: 21383012-21383017 on Build GRCh38
rs867448189 Chr.13: 21382960-21382960 on Build GRCh38
rs1015174093 Chr.13: 21382872-21382872 on Build GRCh38
rs765443059 Chr.13: 21383040-21383040 on Build GRCh38
rs745707311 Chr.13: 21382901-21382901 on Build GRCh38
rs982520120 Chr.13: 21382922-21382922 on Build GRCh38
rs548275879 Chr.13: 21382958-21382958 on Build GRCh38
rs761165273 Chr.13: 21383012-21383012 on Build GRCh38
rs915834555 Chr.13: 21383008-21383008 on Build GRCh38
rs772835851 Chr.13: 21382989-21382989 on Build GRCh38
rs752865074 Chr.13: 21383042-21383042 on Build GRCh38
rs866768875 Chr.13: 21382933-21382933 on Build GRCh38
rs1030491282 Chr.13: 21382902-21382902 on Build GRCh38
rs749120402 Chr.13: 21382931-21382931 on Build GRCh38
rs754384830 Chr.13: 21383027-21383027 on Build GRCh38
rs866432614 Chr.13: 21382920-21382920 on Build GRCh38
rs758529622 Chr.13: 21383044-21383044 on Build GRCh38
rs768463546 Chr.13: 21383032-21383032 on Build GRCh38
rs375991719 Chr.13: 21382892-21382892 on Build GRCh38
rs955124159 Chr.13: 21382912-21382912 on Build GRCh38
rs866796461 Chr.13: 21383006-21383006 on Build GRCh38
rs138594651 Chr.13: 21382981-21382981 on Build GRCh38
rs938620517 Chr.13: 21383034-21383034 on Build GRCh38
rs773810335 Chr.13: 21382980-21382980 on Build GRCh38
rs775500832 Chr.13: 21382928-21382928 on Build GRCh38
rs769464750 Chr.13: 21382919-21382919 on Build GRCh38
rs551868346 Chr.13: 21382984-21382984 on Build GRCh38
rs766589429 Chr.13: 21383018-21383018 on Build GRCh38
rs758657229 Chr.13: 21382885-21382885 on Build GRCh38
rs540987642 Chr.13: 21382938-21382938 on Build GRCh38

View Full Product Details