Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 31989480-31989741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF860
Entrez Gene ID
344787
Gene Name
zinc finger protein 860
Gene Aliases
-
UniGene
Hs.585139
Gene Chromosome Location
Chr.3: 31981774-32005112 on Build GRCh38
Transcripts
NM_001137674.2 XM_017006299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7193875 c.560C>G p.S187C Chr.3: 31989639-31989639 on Build GRCh38
149347 c.428G>A p.R143Q Chr.3: 31989507-31989507 on Build GRCh38
2983951 c.595C>T p.P199S Chr.3: 31989674-31989674 on Build GRCh38
1495507 c.564A>G p.S188S Chr.3: 31989643-31989643 on Build GRCh38
6218192 c.597C>T p.P199P Chr.3: 31989676-31989676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759947074 Chr.3: 31989523-31989523 on Build GRCh38
rs750571902 Chr.3: 31989564-31989564 on Build GRCh38
rs752476083 Chr.3: 31989680-31989680 on Build GRCh38
rs750707145 Chr.3: 31989666-31989666 on Build GRCh38
rs878904354 Chr.3: 31989581-31989581 on Build GRCh38
rs140232911 Chr.3: 31989561-31989561 on Build GRCh38
rs765183438 Chr.3: 31989663-31989663 on Build GRCh38
rs756039556 Chr.3: 31989590-31989590 on Build GRCh38
rs4639011 Chr.3: 31989506-31989506 on Build GRCh38
rs776939738 Chr.3: 31989651-31989651 on Build GRCh38
rs957241016 Chr.3: 31989600-31989600 on Build GRCh38
rs991328848 Chr.3: 31989613-31989613 on Build GRCh38
rs201604355 Chr.3: 31989631-31989631 on Build GRCh38
rs772678376 Chr.3: 31989503-31989503 on Build GRCh38
rs764339985 Chr.3: 31989536-31989536 on Build GRCh38
rs772610191 Chr.3: 31989638-31989638 on Build GRCh38
rs374051878 Chr.3: 31989592-31989592 on Build GRCh38
rs775996535 Chr.3: 31989642-31989642 on Build GRCh38
rs570687729 Chr.3: 31989507-31989507 on Build GRCh38
rs773815563 Chr.3: 31989514-31989514 on Build GRCh38
rs774082577 Chr.3: 31989632-31989632 on Build GRCh38
rs376800921 Chr.3: 31989562-31989562 on Build GRCh38
rs535280784 Chr.3: 31989576-31989576 on Build GRCh38
rs767951119 Chr.3: 31989527-31989527 on Build GRCh38
rs745458067 Chr.3: 31989636-31989636 on Build GRCh38
rs553818646 Chr.3: 31989622-31989622 on Build GRCh38
rs4449357 Chr.3: 31989532-31989532 on Build GRCh38
rs755844281 Chr.3: 31989684-31989684 on Build GRCh38
rs368805853 Chr.3: 31989675-31989675 on Build GRCh38
rs749136931 Chr.3: 31989694-31989694 on Build GRCh38
rs776159912 Chr.3: 31989528-31989528 on Build GRCh38
rs576427985 Chr.3: 31989672-31989672 on Build GRCh38
rs906726837 Chr.3: 31989607-31989607 on Build GRCh38
rs6419811 Chr.3: 31989643-31989643 on Build GRCh38
rs777769636 Chr.3: 31989687-31989687 on Build GRCh38
rs769343203 Chr.3: 31989501-31989501 on Build GRCh38
rs145325915 Chr.3: 31989575-31989575 on Build GRCh38
rs201681180 Chr.3: 31989703-31989703 on Build GRCh38
rs187640039 Chr.3: 31989502-31989502 on Build GRCh38
rs753943367 Chr.3: 31989548-31989548 on Build GRCh38
rs111849538 Chr.3: 31989652-31989652 on Build GRCh38

View Full Product Details