Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 225987956-225988217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDE2
Entrez Gene ID
163859
Gene Name
SDE2 telomere maintenance homolog
Gene Aliases
C1orf55, dJ671D7.1
UniGene
Hs.520192
Gene Chromosome Location
Chr.1: 225982702-225999365 on Build GRCh38
Transcripts
NM_152608.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3671638 c.818C>A p.P273Q Chr.1: 225988176-225988176 on Build GRCh38
3671637 c.854C>A p.P285Q Chr.1: 225988176-225988176 on Build GRCh38
5845639 c.1073C>T p.A358V Chr.1: 225987957-225987957 on Build GRCh38
5155175 c.905C>T p.T302I Chr.1: 225988089-225988089 on Build GRCh38
679369 c.826G>C p.D276H Chr.1: 225988168-225988168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776219134 Chr.1: 225988067-225988067 on Build GRCh38
rs34348128 Chr.1: 225988094-225988094 on Build GRCh38
rs765075570 Chr.1: 225988026-225988026 on Build GRCh38
rs746943280 Chr.1: 225988159-225988159 on Build GRCh38
rs752367074 Chr.1: 225988088-225988088 on Build GRCh38
rs369143013 Chr.1: 225988185-225988185 on Build GRCh38
rs779041650 Chr.1: 225988132-225988132 on Build GRCh38
rs773310192 Chr.1: 225988054-225988054 on Build GRCh38
rs752902169 Chr.1: 225988175-225988175 on Build GRCh38
rs532873318 Chr.1: 225988033-225988033 on Build GRCh38
rs756546478 Chr.1: 225988114-225988114 on Build GRCh38
rs751138203 Chr.1: 225988193-225988193 on Build GRCh38
rs761083095 Chr.1: 225988061-225988063 on Build GRCh38
rs769595056 Chr.1: 225988176-225988176 on Build GRCh38
rs780607277 Chr.1: 225988116-225988116 on Build GRCh38
rs772298701 Chr.1: 225988146-225988146 on Build GRCh38
rs375197716 Chr.1: 225988162-225988162 on Build GRCh38
rs746335165 Chr.1: 225988021-225988021 on Build GRCh38
rs112422303 Chr.1: 225988042-225988042 on Build GRCh38
rs756495190 Chr.1: 225988027-225988027 on Build GRCh38
rs759366781 Chr.1: 225988071-225988071 on Build GRCh38
rs748358175 Chr.1: 225988140-225988140 on Build GRCh38
rs551763102 Chr.1: 225988120-225988120 on Build GRCh38
rs771053978 Chr.1: 225988160-225988160 on Build GRCh38
rs190593519 Chr.1: 225988029-225988029 on Build GRCh38
rs771734154 Chr.1: 225987977-225987977 on Build GRCh38
rs780904838 Chr.1: 225988060-225988060 on Build GRCh38
rs61731742 Chr.1: 225988089-225988089 on Build GRCh38
rs113721886 Chr.1: 225988062-225988062 on Build GRCh38
rs763599065 Chr.1: 225988186-225988186 on Build GRCh38
rs1027701712 Chr.1: 225988142-225988142 on Build GRCh38
rs1018922837 Chr.1: 225988093-225988093 on Build GRCh38
rs781696941 Chr.1: 225988002-225988002 on Build GRCh38
rs768827130 Chr.1: 225988030-225988030 on Build GRCh38
rs773363376 Chr.1: 225988152-225988152 on Build GRCh38
rs775028741 Chr.1: 225988181-225988181 on Build GRCh38
rs1051113051 Chr.1: 225988082-225988082 on Build GRCh38
rs752211409 Chr.1: 225987988-225987988 on Build GRCh38
rs993511554 Chr.1: 225988113-225988113 on Build GRCh38
rs776612834 Chr.1: 225988164-225988164 on Build GRCh38
rs757906431 Chr.1: 225988000-225988000 on Build GRCh38
rs368278911 Chr.1: 225988108-225988108 on Build GRCh38
rs372025839 Chr.1: 225988129-225988129 on Build GRCh38
rs61998244 Chr.1: 225988144-225988144 on Build GRCh38
rs754370563 Chr.1: 225988138-225988138 on Build GRCh38
rs764500509 Chr.1: 225988067-225988069 on Build GRCh38
rs748224059 Chr.1: 225988037-225988037 on Build GRCh38

View Full Product Details