Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 99435735-99435962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSTD3
Entrez Gene ID
100130890
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 3
Gene Aliases
-
UniGene
Hs.634506
Gene Chromosome Location
Chr.6: 99424854-99579288 on Build GRCh38
Transcripts
XM_017010141.1 NM_001080481.1 XM_005267169.3 XM_005267170.4 XM_005267172.3 XM_011536200.2 XM_017011381.1 XM_017011382.1 XM_017011383.1 XM_017011384.1

Gene Symbol
USP45
Entrez Gene ID
85015
Gene Name
ubiquitin specific peptidase 45
Gene Aliases
-
UniGene
Hs.143410
Gene Chromosome Location
Chr.6: 99432306-99515770 on Build GRCh38
Transcripts
XM_017010141.1 NM_001080481.1 XM_005267169.3 XM_005267170.4 XM_005267172.3 XM_011536200.2 XM_017011381.1 XM_017011382.1 XM_017011383.1 XM_017011384.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6300341 c.2324C>T p.A775V Chr.6: 99435837-99435837 on Build GRCh38
1082811 c.2422C>A p.L808I Chr.6: 99435739-99435739 on Build GRCh38
1082812 c.2360T>G p.V787G Chr.6: 99435801-99435801 on Build GRCh38
303534 c.2316T>C p.G772G Chr.6: 99435845-99435845 on Build GRCh38
6012412 c.2319G>C p.L773F Chr.6: 99435842-99435842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535792153 Chr.6: 99435826-99435826 on Build GRCh38
rs779774399 Chr.6: 99435860-99435860 on Build GRCh38
rs759068531 Chr.6: 99435924-99435924 on Build GRCh38
rs764445412 Chr.6: 99435836-99435836 on Build GRCh38
rs776643437 Chr.6: 99435813-99435813 on Build GRCh38
rs756461122 Chr.6: 99435784-99435787 on Build GRCh38
rs766700763 Chr.6: 99435857-99435862 on Build GRCh38
rs978274582 Chr.6: 99435868-99435868 on Build GRCh38
rs375582786 Chr.6: 99435837-99435837 on Build GRCh38
rs778464568 Chr.6: 99435764-99435764 on Build GRCh38
rs749973197 Chr.6: 99435841-99435841 on Build GRCh38
rs745503768 Chr.6: 99435801-99435801 on Build GRCh38
rs761118434 Chr.6: 99435832-99435832 on Build GRCh38
rs1010937705 Chr.6: 99435931-99435931 on Build GRCh38
rs1028262889 Chr.6: 99435900-99435900 on Build GRCh38
rs746847692 Chr.6: 99435805-99435805 on Build GRCh38
rs9402791 Chr.6: 99435818-99435818 on Build GRCh38
rs889813334 Chr.6: 99435904-99435904 on Build GRCh38
rs6570065 Chr.6: 99435828-99435828 on Build GRCh38
rs112515167 Chr.6: 99435772-99435772 on Build GRCh38
rs753968070 Chr.6: 99435860-99435862 on Build GRCh38
rs886273945 Chr.6: 99435783-99435783 on Build GRCh38
rs111350916 Chr.6: 99435780-99435780 on Build GRCh38
rs768428664 Chr.6: 99435812-99435812 on Build GRCh38
rs550114518 Chr.6: 99435802-99435802 on Build GRCh38
rs539929428 Chr.6: 99435926-99435926 on Build GRCh38
rs779789728 Chr.6: 99435804-99435804 on Build GRCh38
rs946979224 Chr.6: 99435859-99435859 on Build GRCh38
rs541374637 Chr.6: 99435861-99435862 on Build GRCh38
rs565974686 Chr.6: 99435885-99435885 on Build GRCh38
rs749258404 Chr.6: 99435816-99435816 on Build GRCh38
rs372586446 Chr.6: 99435830-99435830 on Build GRCh38
rs758003014 Chr.6: 99435850-99435850 on Build GRCh38

View Full Product Details