Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50518100-50518329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAPH2
Entrez Gene ID
29781
Gene Name
non-SMC condensin II complex subunit H2
Gene Aliases
CAPH2
UniGene
Hs.730607
Gene Chromosome Location
Chr.22: 50508216-50524780 on Build GRCh38
Transcripts
NM_001185011.1 NM_014551.4 NM_152299.3 XM_005261912.4 XM_011530685.2 XM_017028793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4503431 c.639C>T p.T213T Chr.22: 50518271-50518271 on Build GRCh38
5745506 c.599_600insC p.M202fs*20 Chr.22: 50518231-50518232 on Build GRCh38
6722793 c.549G>A p.T183T Chr.22: 50518181-50518181 on Build GRCh38
4105206 c.553G>A p.V185I Chr.22: 50518185-50518185 on Build GRCh38
1154045 c.614C>T p.A205V Chr.22: 50518246-50518246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765913247 Chr.22: 50518250-50518250 on Build GRCh38
rs757866198 Chr.22: 50518180-50518180 on Build GRCh38
rs762771405 Chr.22: 50518301-50518301 on Build GRCh38
rs749942415 Chr.22: 50518178-50518178 on Build GRCh38
rs764830899 Chr.22: 50518232-50518232 on Build GRCh38
rs756300446 Chr.22: 50518206-50518206 on Build GRCh38
rs764886951 Chr.22: 50518175-50518175 on Build GRCh38
rs749407343 Chr.22: 50518214-50518214 on Build GRCh38
rs781476433 Chr.22: 50518187-50518187 on Build GRCh38
rs996478861 Chr.22: 50518146-50518146 on Build GRCh38
rs148754615 Chr.22: 50518183-50518183 on Build GRCh38
rs147275808 Chr.22: 50518138-50518138 on Build GRCh38
rs761602714 Chr.22: 50518229-50518229 on Build GRCh38
rs760671058 Chr.22: 50518140-50518140 on Build GRCh38
rs1059500 Chr.22: 50518223-50518223 on Build GRCh38
rs763837851 Chr.22: 50518151-50518151 on Build GRCh38
rs747919278 Chr.22: 50518304-50518304 on Build GRCh38
rs771186931 Chr.22: 50518215-50518215 on Build GRCh38
rs143114125 Chr.22: 50518247-50518247 on Build GRCh38
rs768483694 Chr.22: 50518296-50518296 on Build GRCh38
rs755495949 Chr.22: 50518263-50518263 on Build GRCh38
rs745560670 Chr.22: 50518135-50518135 on Build GRCh38
rs776303299 Chr.22: 50518225-50518225 on Build GRCh38
rs753004631 Chr.22: 50518273-50518273 on Build GRCh38
rs776485524 Chr.22: 50518163-50518163 on Build GRCh38
rs1136707 Chr.22: 50518277-50518277 on Build GRCh38
rs150885852 Chr.22: 50518202-50518202 on Build GRCh38
rs139403039 Chr.22: 50518246-50518246 on Build GRCh38
rs141520277 Chr.22: 50518185-50518185 on Build GRCh38
rs201984002 Chr.22: 50518295-50518295 on Build GRCh38
rs756457575 Chr.22: 50518281-50518281 on Build GRCh38
rs747014361 Chr.22: 50518222-50518222 on Build GRCh38
rs774544738 Chr.22: 50518218-50518218 on Build GRCh38
rs778071708 Chr.22: 50518208-50518208 on Build GRCh38
rs761431324 Chr.22: 50518171-50518171 on Build GRCh38
rs768006797 Chr.22: 50518265-50518265 on Build GRCh38
rs201367371 Chr.22: 50518181-50518181 on Build GRCh38
rs139219191 Chr.22: 50518255-50518255 on Build GRCh38
rs770050043 Chr.22: 50518128-50518128 on Build GRCh38
rs778161451 Chr.22: 50518293-50518293 on Build GRCh38
rs971752138 Chr.22: 50518248-50518248 on Build GRCh38
rs376724692 Chr.22: 50518302-50518302 on Build GRCh38
rs772137492 Chr.22: 50518137-50518137 on Build GRCh38
rs200728457 Chr.22: 50518251-50518251 on Build GRCh38
rs199873835 Chr.22: 50518294-50518294 on Build GRCh38
rs773220074 Chr.22: 50518134-50518134 on Build GRCh38
rs371503348 Chr.22: 50518224-50518224 on Build GRCh38

View Full Product Details