Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 131381896-131382121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT16
Entrez Gene ID
131870
Gene Name
nudix hydrolase 16
Gene Aliases
-
UniGene
Hs.282050
Gene Chromosome Location
Chr.3: 131381671-131388830 on Build GRCh38
Transcripts
NM_001171905.1 NM_001171906.1 NM_152395.2 NR_033268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5677339 c.112C>G p.R38G Chr.3: 131381916-131381916 on Build GRCh38
7380669 c.67C>G p.L23V Chr.3: 131382112-131382112 on Build GRCh38
7128561 c.186G>A p.V62V Chr.3: 131382093-131382093 on Build GRCh38
374468 c.14G>A p.R5H Chr.3: 131381917-131381917 on Build GRCh38
6281371 c.52A>T p.T18S Chr.3: 131382097-131382097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747285916 Chr.3: 131382042-131382042 on Build GRCh38
rs773350182 Chr.3: 131381931-131381931 on Build GRCh38
rs756770071 Chr.3: 131381922-131381922 on Build GRCh38
rs763858616 Chr.3: 131382010-131382010 on Build GRCh38
rs746029308 Chr.3: 131382060-131382060 on Build GRCh38
rs1040814300 Chr.3: 131382099-131382099 on Build GRCh38
rs943763741 Chr.3: 131382083-131382083 on Build GRCh38
rs768544264 Chr.3: 131382084-131382084 on Build GRCh38
rs749950474 Chr.3: 131382029-131382029 on Build GRCh38
rs781055611 Chr.3: 131381937-131381937 on Build GRCh38
rs141462594 Chr.3: 131382065-131382065 on Build GRCh38
rs767264384 Chr.3: 131382098-131382098 on Build GRCh38
rs761561572 Chr.3: 131382014-131382014 on Build GRCh38
rs748586686 Chr.3: 131381952-131381952 on Build GRCh38
rs775402238 Chr.3: 131381995-131381995 on Build GRCh38
rs763803559 Chr.3: 131381917-131381917 on Build GRCh38
rs762795361 Chr.3: 131382009-131382009 on Build GRCh38
rs989467037 Chr.3: 131381935-131381935 on Build GRCh38
rs375496917 Chr.3: 131381953-131381953 on Build GRCh38
rs751215085 Chr.3: 131382013-131382013 on Build GRCh38
rs1037253368 Chr.3: 131381945-131381945 on Build GRCh38
rs759475591 Chr.3: 131381985-131381985 on Build GRCh38
rs779617762 Chr.3: 131382035-131382035 on Build GRCh38
rs776792947 Chr.3: 131381967-131381967 on Build GRCh38
rs369569819 Chr.3: 131381921-131381921 on Build GRCh38
rs771018348 Chr.3: 131381966-131381966 on Build GRCh38
rs940152949 Chr.3: 131381944-131381944 on Build GRCh38
rs527720651 Chr.3: 131382049-131382049 on Build GRCh38
rs753230762 Chr.3: 131382036-131382036 on Build GRCh38
rs755534807 Chr.3: 131381933-131381933 on Build GRCh38
rs755572495 Chr.3: 131382034-131382034 on Build GRCh38
rs769728270 Chr.3: 131381989-131381989 on Build GRCh38
rs978220562 Chr.3: 131382012-131382012 on Build GRCh38
rs780725909 Chr.3: 131381928-131381928 on Build GRCh38
rs376932370 Chr.3: 131382097-131382097 on Build GRCh38
rs774219967 Chr.3: 131382093-131382093 on Build GRCh38
rs148362533 Chr.3: 131382045-131382045 on Build GRCh38
rs767031967 Chr.3: 131382025-131382025 on Build GRCh38
rs1000317911 Chr.3: 131382069-131382069 on Build GRCh38
rs756149094 Chr.3: 131381951-131381951 on Build GRCh38
rs773614501 Chr.3: 131381960-131381960 on Build GRCh38
rs557974832 Chr.3: 131381975-131381975 on Build GRCh38
rs369735356 Chr.3: 131381962-131381962 on Build GRCh38
rs762850285 Chr.3: 131382080-131382080 on Build GRCh38
rs910091334 Chr.3: 131381943-131381943 on Build GRCh38
rs767675204 Chr.3: 131381964-131381983 on Build GRCh38
rs372691131 Chr.3: 131382037-131382037 on Build GRCh38

View Full Product Details