Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49061327-49061572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTF4
Entrez Gene ID
4909
Gene Name
neurotrophin 4
Gene Aliases
GLC10, GLC1O, NT-4, NT-4/5, NT-5, NT4, NT5, NTF5
UniGene
Hs.266902
Gene Chromosome Location
Chr.19: 49058284-49065076 on Build GRCh38
Transcripts
NM_006179.4 XM_005258962.3 XM_006723232.3 XM_011527008.2 XM_011527009.2 XM_011527010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2752011 c.626G>A p.R209Q Chr.19: 49061372-49061372 on Build GRCh38
6403508 c.565G>T p.G189C Chr.19: 49061433-49061433 on Build GRCh38
4624056 c.477G>A p.R159R Chr.19: 49061521-49061521 on Build GRCh38
4400453 c.580C>T p.R194* Chr.19: 49061418-49061418 on Build GRCh38
1286708 c.474C>T p.C158C Chr.19: 49061524-49061524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533615274 Chr.19: 49061426-49061426 on Build GRCh38
rs776012757 Chr.19: 49061469-49061469 on Build GRCh38
rs200878406 Chr.19: 49061499-49061499 on Build GRCh38
rs777832037 Chr.19: 49061370-49061370 on Build GRCh38
rs776177618 Chr.19: 49061482-49061482 on Build GRCh38
rs765132008 Chr.19: 49061401-49061401 on Build GRCh38
rs562474394 Chr.19: 49061372-49061372 on Build GRCh38
rs759034173 Chr.19: 49061540-49061540 on Build GRCh38
rs374345134 Chr.19: 49061545-49061545 on Build GRCh38
rs772499451 Chr.19: 49061457-49061457 on Build GRCh38
rs369066724 Chr.19: 49061517-49061517 on Build GRCh38
rs749392406 Chr.19: 49061438-49061438 on Build GRCh38
rs769822264 Chr.19: 49061369-49061369 on Build GRCh38
rs142027939 Chr.19: 49061437-49061437 on Build GRCh38
rs746321607 Chr.19: 49061538-49061538 on Build GRCh38
rs779851034 Chr.19: 49061414-49061414 on Build GRCh38
rs200675509 Chr.19: 49061373-49061373 on Build GRCh38
rs200081067 Chr.19: 49061400-49061400 on Build GRCh38
rs797044939 Chr.19: 49061477-49061477 on Build GRCh38
rs764829441 Chr.19: 49061486-49061486 on Build GRCh38
rs768336179 Chr.19: 49061546-49061546 on Build GRCh38
rs781438973 Chr.19: 49061358-49061358 on Build GRCh38
rs772979004 Chr.19: 49061503-49061503 on Build GRCh38
rs746640305 Chr.19: 49061514-49061514 on Build GRCh38
rs375526828 Chr.19: 49061433-49061433 on Build GRCh38
rs766101589 Chr.19: 49061509-49061509 on Build GRCh38
rs121918427 Chr.19: 49061382-49061382 on Build GRCh38
rs763358860 Chr.19: 49061493-49061494 on Build GRCh38
rs780565192 Chr.19: 49061541-49061541 on Build GRCh38
rs1025529881 Chr.19: 49061417-49061417 on Build GRCh38
rs748387322 Chr.19: 49061362-49061362 on Build GRCh38
rs373152998 Chr.19: 49061393-49061393 on Build GRCh38
rs1035055452 Chr.19: 49061385-49061385 on Build GRCh38
rs377015369 Chr.19: 49061395-49061395 on Build GRCh38
rs572957948 Chr.19: 49061408-49061408 on Build GRCh38
rs775964511 Chr.19: 49061465-49061466 on Build GRCh38
rs369788069 Chr.19: 49061418-49061418 on Build GRCh38
rs371734511 Chr.19: 49061505-49061505 on Build GRCh38
rs761282348 Chr.19: 49061484-49061484 on Build GRCh38
rs865923726 Chr.19: 49061500-49061500 on Build GRCh38
rs779054536 Chr.19: 49061537-49061537 on Build GRCh38
rs922351065 Chr.19: 49061519-49061519 on Build GRCh38
rs371861346 Chr.19: 49061378-49061378 on Build GRCh38
rs370717457 Chr.19: 49061466-49061466 on Build GRCh38
rs764831671 Chr.19: 49061493-49061493 on Build GRCh38
rs147294437 Chr.19: 49061446-49061446 on Build GRCh38
rs555228417 Chr.19: 49061352-49061352 on Build GRCh38
rs750214028 Chr.19: 49061405-49061405 on Build GRCh38
rs367957078 Chr.19: 49061481-49061481 on Build GRCh38
rs374415552 Chr.19: 49061471-49061471 on Build GRCh38
rs758136156 Chr.19: 49061411-49061411 on Build GRCh38
rs764263332 Chr.19: 49061523-49061523 on Build GRCh38
rs753931867 Chr.19: 49061527-49061527 on Build GRCh38
rs975549134 Chr.19: 49061515-49061515 on Build GRCh38
rs757244498 Chr.19: 49061535-49061535 on Build GRCh38
rs760409652 Chr.19: 49061390-49061390 on Build GRCh38
rs746418406 Chr.19: 49061453-49061453 on Build GRCh38
rs779336456 Chr.19: 49061441-49061441 on Build GRCh38
rs1033709780 Chr.19: 49061476-49061476 on Build GRCh38
rs974105707 Chr.19: 49061455-49061455 on Build GRCh38
rs144346396 Chr.19: 49061439-49061439 on Build GRCh38
rs933661685 Chr.19: 49061531-49061531 on Build GRCh38
rs121918428 Chr.19: 49061381-49061381 on Build GRCh38
rs755200724 Chr.19: 49061356-49061356 on Build GRCh38

View Full Product Details