Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73634603-73634830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOZ1
Entrez Gene ID
58529
Gene Name
myozenin 1
Gene Aliases
CS-2, FATZ, MYOZ
UniGene
Hs.238756
Gene Chromosome Location
Chr.10: 73631612-73641757 on Build GRCh38
Transcripts
NM_021245.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4015770 c.315C>A p.F105L Chr.10: 73634671-73634671 on Build GRCh38
4614633 c.287_288insG p.Q97fs*34 Chr.10: 73634698-73634699 on Build GRCh38
4015769 c.344G>A p.G115D Chr.10: 73634642-73634642 on Build GRCh38
5796624 c.360C>A p.G120G Chr.10: 73634626-73634626 on Build GRCh38
5688395 c.292C>T p.L98L Chr.10: 73634694-73634694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751483817 Chr.10: 73634644-73634644 on Build GRCh38
rs767385902 Chr.10: 73634732-73634732 on Build GRCh38
rs1011395810 Chr.10: 73634783-73634783 on Build GRCh38
rs921632302 Chr.10: 73634702-73634702 on Build GRCh38
rs758824974 Chr.10: 73634687-73634687 on Build GRCh38
rs144795054 Chr.10: 73634704-73634704 on Build GRCh38
rs746286638 Chr.10: 73634681-73634681 on Build GRCh38
rs370996035 Chr.10: 73634692-73634692 on Build GRCh38
rs772911864 Chr.10: 73634705-73634705 on Build GRCh38
rs762712605 Chr.10: 73634708-73634708 on Build GRCh38
rs202128209 Chr.10: 73634669-73634669 on Build GRCh38
rs768785792 Chr.10: 73634698-73634699 on Build GRCh38
rs188206000 Chr.10: 73634753-73634753 on Build GRCh38
rs764127123 Chr.10: 73634652-73634652 on Build GRCh38
rs538058320 Chr.10: 73634698-73634698 on Build GRCh38
rs372939816 Chr.10: 73634716-73634716 on Build GRCh38
rs780759330 Chr.10: 73634689-73634689 on Build GRCh38
rs772893553 Chr.10: 73634630-73634630 on Build GRCh38
rs750639897 Chr.10: 73634774-73634774 on Build GRCh38
rs142497147 Chr.10: 73634758-73634758 on Build GRCh38
rs139908512 Chr.10: 73634643-73634643 on Build GRCh38
rs759500912 Chr.10: 73634725-73634725 on Build GRCh38
rs926025564 Chr.10: 73634776-73634776 on Build GRCh38
rs753905596 Chr.10: 73634653-73634653 on Build GRCh38
rs765437792 Chr.10: 73634765-73634765 on Build GRCh38
rs766138458 Chr.10: 73634711-73634711 on Build GRCh38
rs776022781 Chr.10: 73634696-73634696 on Build GRCh38

View Full Product Details